EMAIL THIS PAGE TO A FRIEND
Journal of medicinal food
Osmanthus fragrans Flower Extract and Acteoside Protect Against d-Galactose-Induced Aging in an ICR Mouse Model.
PMID 26181905
Abstract
Osmanthus fragrans flower extract (OFE) is an organic extract from O. fragrans flower, which exhibits neuroprotective, free radical scavenging, and antioxidant effects. Therefore, the protective effect of OFE and acteoside against aging was studied. An aging ICR mouse model was established by chronically administering d-galactose (250 mg/kg) for 8 weeks. d-galactose induced spatial learning and memory impairments that were successfully inhibited by OFE and acteoside, which could shorten escape latency, improve platform crossing times, and increase zone time. The antioxidant potential of OFE and acteoside in vivo was evaluated by estimating the following: activities of antioxidant enzymes, such as glutathione peroxidase and aging-related enzyme, particularly monoamine oxidase; contents of lipid peroxidation methane dicarboxylic aldehyde, advanced glycation end products, and 8-hydroxy-2'-deoxyguanosine (a DNA damage product); and levels of nuclear factor-erythroid 2-related factor 2. OFE and acteoside also inhibited d-galactose-induced neurological aging by suppressing the increase in glial fibrillary acidic protein and neurotrophin-3. Considering the dose-dependent protective effects of OFE and acteoside, we concluded that OFE, rich in acteoside, was a good source of natural antiaging compounds.