EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. A
Sensitive and direct determination of lithium by mixed-mode chromatography and charged aerosol detection.
PMID 26184708
Abstract
A sensitive analytical method using mixed mode HPLC separation coupled with charged aerosol detection (CAD) was developed for quantitative analysis of lithium. The method is capable of separating lithium ion from different drug matrices and other ions in a single run thus eliminating the organic matrix and ionic analyte interferences without extensive sample preparation such as derivatization and extraction. The separation space and chromatographic conditions are defined by systematic studies of the retention behaviors of lithium and potential interfering ions and different type of pharmaceutical APIs (active pharmaceutical ingredients) under reversed-phase, HILIC and cation/anion exchange mechanisms. Compared to other current analytical techniques for lithium analysis, the presented method provides a new approach and demonstrates high sensitivity (0.02ng for LOD and 0.08ng for LOQ in both standard and sample solution). The method has been validated for pharmaceutical samples and can be potentially applied to biological, food and environmental samples.