EMAIL THIS PAGE TO A FRIEND
Dalton transactions (Cambridge, England : 2003)
Synthesis and characterisation of halide, separated ion pair, and hydride cyclopentadienyl iron bis(diphenylphosphino)ethane derivatives.
PMID 26185807
Abstract
Treatment of anhydrous FeX2 (X = Cl, Br, I) with one equivalent of bis(diphenylphosphino)ethane (dppe) in refluxing THF afforded analytically pure white (X = Cl), light green (X = Br), and yellow (X = I) [FeX2(dppe)]n (X = Cl, ; Br, ; I, ). Complexes are excellent synthons from which to prepare a range of cyclopentadienyl derivatives. Specifically, treatment of with alkali metal salts of C5H5 (Cp, series ), C5Me5 (Cp*, series ), C5H4SiMe3 (Cp', series ), C5H3(SiMe3)2 (Cp'', series ), and C5H3(Bu(t))2 (Cp(tt), series ) afforded [Fe(Cp(†))(Cl)(dppe)] , [Fe(Cp(†))(Br)(dppe)] , and [Fe(Cp(†))(I)(dppe)] (Cp(†) = Cp, Cp*, Cp', Cp'', or Cp(tt)). Dissolution of in acetonitrile, or treatment of with Me3SiI in acetonitrile (no halide exchange reactions were observed in other solvents) afforded the separated ion pair complexes [Fe(Cp(†))(NCMe)(dppe)][I] . Attempts to reduce , , and with a variety of reductants (Li-Cs, KC8, Na/Hg) were unsuccessful. Treatment of with LiAlH4 gave the hydride derivatives [Fe(Cp(†))(H)(dppe)] . This report provides a systematic account of reliable methods of preparing these complexes which may find utility in molecular wire and metal-metal bond chemistries. The complexes reported herein have been characterised by X-ray diffraction, NMR, IR, UV/Vis, and Mössbauer spectroscopies, cyclic voltammetry, density functional theory calculations, and elemental analyses, which have enabled us to elucidate the electronic structure of the complexes and probe the variation of iron redox properties as a function of varying the cyclopentadienyl or halide ligand.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
357340
Rhodium, foil, thickness 0.025 mm, 99.9% trace metals basis
Rh
204218
Rhodium, powder, 99.95% trace metals basis
Rh
267368
Rhodium, sponge, 99.9% trace metals basis
Rh
277916
Rhodium, wire, diam. 1.0 mm, 99.9% trace metals basis
Rh
GF94930667
Rhodium, microfoil, disks, 10mm, thinness 0.25μm, specific density 310μg/cm2, permanent mylar 3.5μm support, 99.99%
Rh
GF64213680
Rhodium, wire reel, 0.05m, diameter 0.12mm, as drawn, 99.9%
Rh
GF37380089
Rhodium, wire reel, 0.2m, diameter 0.25mm, hard, 99.9%
Rh
GF45819995
Rhodium, wire reel, 0.05m, diameter 0.25mm, hard, 99.9%
Rh
GF83497652
Rhodium, wire reel, 0.05m, diameter 1.0mm, as drawn, 99.9%
Rh
GF26710680
Rhodium, wire reel, 0.1m, diameter 0.12mm, as drawn, 99.9%
Rh
GF27499045
Rhodium, wire reel, 0.1m, diameter 0.25mm, hard, 99.9%
Rh
GF06539979
Rhodium, wire reel, 0.1m, diameter 0.5mm, as drawn, 99.9%
Rh
GF37207166
Rhodium, wire reel, 0.1m, diameter 1.0mm, as drawn, 99.9%
Rh
GF38835737
Rhodium, wire reel, 0.2m, diameter 0.12mm, as drawn, 99.9%
Rh
GF17496973
Rhodium, wire reel, 0.2m, diameter 0.5mm, as drawn, 99.9%
Rh
GF54466937
Rhodium, wire reel, 0.5m, diameter 0.12mm, as drawn, 99.9%
Rh
GF72510364
Rhodium, wire reel, 0.5m, diameter 0.25mm, hard, 99.9%
Rh
GF44569907
Rhodium, foil, 25x25mm, thickness 0.125mm, as rolled, 99.9%
Rh
GF80841146
Rhodium, foil, 25x25mm, thickness 0.15mm, as rolled, 99.9%
Rh
GF40539405
Rhodium, foil, 25x25mm, thickness 0.1mm, as rolled, 99.9%
Rh
GF20569697
Rhodium, foil, 12mm disks, thickness 0.5mm, as rolled, 99.9%
Rh
GF61934429
Rhodium, foil, 12mm disks, thickness 1.0mm, as rolled, 99.9%
Rh
GF36379345
Rhodium, foil, light tested, 15x15mm, thickness 0.05mm, as rolled, 99.9%
Rh
GF37647142
Rhodium, foil, light tested, 25x25mm, thickness 0.025mm, as rolled, x-ray quality, 99.9%
Rh
GF57675561
Rhodium, foil, light tested, 25x25mm, thickness 0.05mm, as rolled, 99.9%
Rh
GF12568013
Rhodium, foil, light tested, 25x25mm, thickness 0.05mm, as rolled, x-ray quality, 99.9%
Rh
GF96356559
Rhodium, foil, light tested, 25x25mm, thickness 0.075mm, as rolled, 99.9%
Rh
GF04978963
Rhodium, foil, not light tested, 10x10mm, thickness 0.012mm, as rolled, 99.9%
Rh
GF17514312
Rhodium, foil, not light tested, 25x25mm, thickness 0.00025mm, permanent polyester support, 99.9%
Rh
GF35442727
Rhodium, foil, not light tested, 25x25mm, thickness 0.0005mm, permanent polyester support, 99.85%
Rh