EMAIL THIS PAGE TO A FRIEND
Inflammatory bowel diseases
Effect of Thalidomide on Clinical Remission in Children and Adolescents with Ulcerative Colitis Refractory to Other Immunosuppressives: Pilot Randomized Clinical Trial.
PMID 26185909
Abstract
In a randomized controlled trial, thalidomide has shown to be effective in refractory Crohn's disease in children. This pilot study aimed at evaluating thalidomide in refractory pediatric ulcerative colitis (UC). Double-blind, placebo-controlled randomized clinical trial on thalidomide 1.5 to 2.5 mg/kg/day in children with active UC despite multiple immunosuppressive treatments. In an open-label extension, nonresponders to placebo received thalidomide for an additional 8 weeks; all responders were followed up for a minimum of 52 weeks. Twenty-six children with refractory UC were randomized to thalidomide or placebo. Clinical remission at week 8 was achieved by significantly more children treated with thalidomide {10/12 (83.3%) versus 2/11 (18.8%); risk ratio, 4.5 (95% confidence interval [CI], 1.2-16.4); P = 0.005; number needed to treat, 1.5}. Of the nonresponders to placebo who were switched to thalidomide, 8 of 11 (72.7%) subsequently reached remission at week 8 (risk ratio, 4.0 [95% CI, 1.1-14.7]; number needed to treat, 2.45; P = 0.01). Clinical remission in the thalidomide group was 135.0 weeks (95% CI, 32-238), compared with 8.0 weeks (95% CI, 2.4-13.6) in the placebo group (P < 0.0001). Cumulative incidence of severe adverse events was 3.1 per 1000 patient-weeks. Peripheral neuropathy and amenorrhea were the most frequent adverse events. In this pilot randomized controlled trial on cases of UC refractory to immunosuppressive therapy, thalidomide compared with placebo resulted in improved clinical remission at 8 weeks of treatment and in longer term maintenance of remission. These findings require replication in larger clinical studies evaluating both thalidomide efficacy and safety.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T144
(±)-Thalidomide, ≥98%, powder
C13H10N2O4