EMAIL THIS PAGE TO A FRIEND
Journal of neurotrauma
Acute Phase Proteins in Cerebrospinal Fluid from Dogs with Naturally-Occurring Spinal Cord Injury.
PMID 26186466
Abstract
Spinal cord injury (SCI) affects thousands of people each year and there are no treatments that dramatically improve clinical outcome. Canine intervertebral disc herniation is a naturally-occurring SCI that has similarities to human injury and can be used as a translational model for evaluating therapeutic interventions. Here, we characterized cerebrospinal fluid (CSF) acute phase proteins (APPs) that have altered expression across a spectrum of neurological disorders, using this canine model system. The concentrations of C-reactive protein (CRP), haptoglobin (Hp), alpha-1-glycoprotein, and serum amyloid A were determined in the CSF of 42 acutely injured dogs, compared with 21 healthy control dogs. Concentrations of APPs also were examined with respect to initial injury severity and motor outcome 42 d post-injury. Hp concentration was significantly higher (p<0.0001) in the CSF of affected dogs, compared with healthy control dogs. Additionally, the concentrations of CRP and Hp were significantly (p=0.0001 and p=0.0079, respectively) and positively associated with CSF total protein concentration. The concentrations of CRP and Hp were significantly higher (p=0.0071 and p=0.0197, respectively) in dogs with severe injury, compared with those with mild-to-moderate SCI, but there was no significant correlation between assessed CSF APP concentrations and 42 d motor outcome. This study demonstrated that CSF APPs were dysregulated in dogs with naturally-occurring SCI and could be used as markers for SCI severity. As Hp was increased following severe SCI and is neuroprotective across a number of model systems, it may represent a viable therapeutic target.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F3886
Fentanyl citrate salt
C22H28N2O · C6H8O7
SML0029
Glycopyrrolate, ≥98% (HPLC)
C19H28NO3 · Br