EMAIL THIS PAGE TO A FRIEND
Die Pharmazie
Protective properties of ginsenoside Rb1 against UV-B radiation-induced oxidative stress in human dermal keratinocytes.
PMID 26189299
Abstract
Ginsenosides, also known as ginseng saponins, are responsible for most pharmacological effect of ginseng. Ginsenoside Rb1 (Rb1) exerts a variety of pharmacological properties, including anti-inflammatory, antistress, anti-aging and anti-neurodegenerative activities. The aim of the present work was to assess the skin anti-photoaging properties of Rb1 in human dermal keratinocyte HaCaT cells. The anti-photoaging activity was evaluated by analyzing the levels of reactive oxygen species (ROS) and matrix metalloproteinases (MMPs) as well as cell viability for HaCaT cells under UV-B irradiation. Rb1 was able to suppress the ROS levels which were elevated under UV-B irradiation, and unable to influence cellular survival in UV-B-irradiated HaCaT cells. Rb1 diminished the enhancement of MMP-2 gelatinolytic activity in conditioned medium, which corresponded with the decreased MMP-2 protein levels in both conditioned medium and cellular lysate prepared from UV-B-irradiated HaCaT cultures. Rb1 could restore the total glutathione (GSH) and superoxide dismutase (SOD) activity diminished in UV-B-irradiated HaCaT cells. Ginsenoside Rb1 possesses skin anti-photoaging properties through scavenging ROS and decreasing MMP-2 levels possibly by enhancing antioxidant activity in keratinocytes under UV-B irradiation.