EMAIL THIS PAGE TO A FRIEND
International journal of clinical and experimental pathology
Hypoxia promotes AMP-activated protein kinase (AMPK) and induces apoptosis in mouse osteoblasts.
PMID 26191182
Abstract
The hypoxic environment around the fracture site develops post the blood flow disruption and leads to osteoblast cell death and further impairs fracture healing. Hypoxia usually leads to the mitochondrial dysfunction and then results in apoptotic cell death. AMPK is ubiquitously expressed and functions as an intracellular fuel sensor by maintaining energy balance, as is potentially activated by hypoxia, ischemia, and ROS, however, the regulatory role of AMPK in hypoxia-induced apoptosis in osteoblasts and in the fracture healing has not been identified. In present study, we firstly determined the apoptosis induction by hypoxia in mouse osteoblastic MC3T3-E1 cells via examining the apoptotic cells and the activation of apoptosis-related molecules, then investigated the activation of AMPK signaling by hypoxia via analyzing the phosphorylation of AMPKα and ACC1, finally we explored the association of the AMPK activation with the hypoxia-induced apoptosis using loss-of-function strategy. Results demonstrated that hypoxia induced apoptosis in MC3T3-E1 cells and activated the AMPK signaling. And the knockdown of AMPK via chemical treatment or RNA interfering significantly decreased the hypoxia-induced apoptosis in MC3T3-E1 cells. Taken together, present study unveiled the regulatory role of AMPK signaling in the hypoxia-induced osteoblast apoptosis.