EMAIL THIS PAGE TO A FRIEND
Archiv der Pharmazie
Synthesis and Evaluation of Chroman-4-One Linked to N-Benzyl Pyridinium Derivatives as New Acetylcholinesterase Inhibitors.
PMID 26192069
Abstract
A novel series of chroman-4-one derivatives containing the N-benzyl pyridinium moiety were designed, synthesized, and evaluated for their acetylcholinesterase (AChE) inhibitory activities. Among the various synthesized compounds, (E)-1-(2,3-dibromobenzyl)-4-((7-ethoxy-4-oxochroman-3-ylidene)methyl)pyridinium bromide (8l) depicted the most potent anti-AChE activity (IC50 = 0.048 μM). In addition, the molecular modeling study allowed us to detect possible binding modes that are in full compliance with the observed results through in vitro experiments.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
122351
4-Chromanone, 97%
C9H8O2