EMAIL THIS PAGE TO A FRIEND
Advanced materials (Deerfield Beach, Fla.)
Design of hydrophilic metal organic framework water adsorbents for heat reallocation.
PMID 26193346
Abstract
A new hydrothermally stable Al polycarboxylate metal-organic framework (MOF) based on a heteroatom bio-derived aromatic spacer is designed through a template-free green synthesis process. It appears that in some test conditions this MOF outperforms the heat reallocation performances of commercial SAPO-34.