PloS one
Light-Dependent Aerobic Methane Oxidation Reduces Methane Emissions from Seasonally Stratified Lakes.
PMID 26193458
Abstract
Lakes are a natural source of methane to the atmosphere and contribute significantly to total emissions compared to the oceans. Controls on methane emissions from lake surfaces, particularly biotic processes within anoxic hypolimnia, are only partially understood. Here we investigated biological methane oxidation in the water column of the seasonally stratified Lake Rotsee. A zone of methane oxidation extending from the oxic/anoxic interface into anoxic waters was identified by chemical profiling of oxygen, methane and δ13C of methane. Incubation experiments with 13C-methane yielded highest oxidation rates within the oxycline, and comparable rates were measured in anoxic waters. Despite predominantly anoxic conditions within the zone of methane oxidation, known groups of anaerobic methanotrophic archaea were conspicuously absent. Instead, aerobic gammaproteobacterial methanotrophs were identified as the active methane oxidizers. In addition, continuous oxidation and maximum rates always occurred under light conditions. These findings, along with the detection of chlorophyll a, suggest that aerobic methane oxidation is tightly coupled to light-dependent photosynthetic oxygen production both at the oxycline and in the anoxic bottom layer. It is likely that this interaction between oxygenic phototrophs and aerobic methanotrophs represents a widespread mechanism by which methane is oxidized in lake water, thus diminishing its release into the atmosphere.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
GF93740384
Carbon - Vitreous, rod, 100mm, diameter 10mm, glassy carbon
C
GF65427587
Carbon - Vitreous, rod, 100mm, diameter 3.0mm, glassy carbon
C
GF05962765
Carbon - Vitreous, rod, 100mm, diameter 5.0mm, glassy carbon
C
GF68195403
Carbon - Vitreous, rod, 100mm, diameter 7.0mm, glassy carbon
C
GF23723423
Carbon - Vitreous, rod, 200mm, diameter 1.0mm, glassy carbon
C
GF91785134
Carbon - Vitreous, rod, 200mm, diameter 10mm, glassy carbon
C
GF78447707
Carbon - Vitreous, rod, 200mm, diameter 3.0mm, glassy carbon
C
GF45345562
Carbon - Vitreous, rod, 200mm, diameter 5.0mm, glassy carbon
C
GF24425805
Carbon - Vitreous, rod, 200mm, diameter 7.0mm, glassy carbon
C
GF93251011
Carbon - Vitreous, rod, 50mm, diameter 1.0mm, glassy carbon
C
GF20176680
Carbon - Vitreous, rod, 5mm, diameter 3.0mm, glassy carbon
C
GF59400233
Carbon - Vitreous, tube, 100mm, outside diameter 10mm, inside diameter 3mm, wall thickness 3.5mm, glassy carbon
C
GF39088648
Carbon - Vitreous, tube, 50mm, outside diameter 10mm, inside diameter 3mm, wall thickness 3.5mm, glassy carbon
C
GF07378476
Carbon - Vitreous, foam, 150x150mm, thickness 2.5mm, bulk density 0.05g/cm3, porosity 96.5%
C
GF39873369
Carbon - Vitreous, foam, 150x150mm, thickness 3.2mm, bulk density 0.05g/cm3, porosity 96.5%
C
GF61342220
Carbon - Vitreous, foam, 150x150mm, 0.05g.cmué, porosity 96.5%, 24 pores/cm
C
GF58042324
Carbon - Vitreous, foam, 275x330mm, thickness 3.2mm, bulk density 0.05g/cm3, porosity 96.5%
C
GF12408713
Carbon - Vitreous, foam, 275x330mm, 0.05g.cmué, porosity 96.5%, 24 pores/cm
C
GF04031132
Carbon - Vitreous, foam, 300x300mm, thickness 20mm, bulk density 0.05g/cm3, porosity 96.5%
C
GF06230922
Carbon - Vitreous, foam, 300x300mm, thickness 30mm, bulk density 0.05g/cm3, porosity 96.5%
C
GF95357526
Carbon - Vitreous, foil, 100x100mm, thickness 1.0mm, glassy carbon
C
GF61808889
Carbon - Vitreous, foil, 100x100mm, thickness 2.0mm, glassy carbon
C
GF09325009
Carbon - Vitreous, foil, 100x100mm, thickness 6.0mm, glassy carbon
C
GF22691426
Carbon - Vitreous, foil, 10x10mm, thickness 1.0mm, glassy carbon
C
GF74135628
Carbon - Vitreous, foil, 10x10mm, thickness 2.0mm, glassy carbon
C
GF99431523
Carbon - Vitreous, foil, 10x10mm, thickness 4.0mm, glassy carbon
C
GF44361457
Carbon - Vitreous, foil, 200x200mm, thickness 1.0mm, glassy carbon
C
GF10355958
Carbon - Vitreous, foil, 25x25mm, thickness 0.5mm, glassy carbon
C
GF89520521
Carbon - Vitreous, foil, 25x25mm, thickness 4.0mm, glassy carbon
C
GF96762864
Carbon - Vitreous, foil, 50x50mm, thickness 1.0mm, glassy carbon
C