EMAIL THIS PAGE TO A FRIEND
Nature communications
Phosphorylation status determines the opposing functions of Smad2/Smad3 as STAT3 cofactors in TH17 differentiation.
PMID 26194464
Abstract
Transforming growth factor-β (TGF-β) and interleukin-6 (IL-6) are the pivotal cytokines to induce IL-17-producing CD4(+) T helper cells (TH17); yet their signalling network remains largely unknown. Here we show that the highly homologous TGF-β receptor-regulated Smads (R-Smads): Smad2 and Smad3 oppositely modify STAT3-induced transcription of IL-17A and retinoic acid receptor-related orphan nuclear receptor, RORγt encoded by Rorc, by acting as a co-activator and co-repressor of STAT3, respectively. Smad2 linker phosphorylated by extracellular signal-regulated kinase (ERK) at the serine 255 residue interacts with STAT3 and p300 to transactivate, whereas carboxy-terminal unphosphorylated Smad3 interacts with STAT3 and protein inhibitor of activated STAT3 (PIAS3) to repress the Rorc and Il17a genes. Our work uncovers carboxy-terminal phosphorylation-independent noncanonical R-Smad-STAT3 signalling network in TH17 differentiation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
W217018
2-Butanone, ≥99.5%, FCC, FG
C4H8O
PRS4877
Anti-IL-17 (ab1) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
PRS4887
Anti-IL-17 (ab2) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2502114
Anti-IL17A antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB2502115
Anti-IL17A antibody produced in goat, affinity isolated antibody, buffered aqueous solution
HPA052258
Anti-IL17A antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4300708
Anti-STAT3 (Ab-705) antibody produced in rabbit, affinity isolated antibody
SAB4300327
Anti-STAT3 (Ab-727) antibody produced in rabbit, affinity isolated antibody
SAB2500993
Anti-STAT3 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB1406487
Anti-STAT3 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA001671
Anti-STAT3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2102317
Anti-STAT3 antibody produced in rabbit, affinity isolated antibody
S5933 Anti-STAT3 antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB2104461
Anti-STAT3 antibody produced in rabbit, affinity isolated antibody
SAB2104912
Anti-STAT3 antibody produced in rabbit, affinity isolated antibody
SAB4502871
Anti-STAT3, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4502872
Anti-STAT3, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4502873
Anti-STAT3, C-Terminal antibody produced in rabbit, affinity isolated antibody
L9026
Lys-Lys-Lys-Lys, ≥95% (TLC)
C24H50N8O5
EHU122051 MISSION® esiRNA, esiRNA human STAT3 (esiRNA1)
EMU026711 MISSION® esiRNA, esiRNA targeting mouse Pias3 (esiRNA1)
EHU050391 MISSION® esiRNA, esiRNA human PIAS3 (esiRNA1)
EMU082481 MISSION® esiRNA, esiRNA targeting mouse Stat3 (esiRNA1)
SAB4700340 Monoclonal Anti-IFNG antibody produced in mouse, clone G-23, purified immunoglobulin, buffered aqueous solution
SAB1402815 Monoclonal Anti-IFNG, (C-terminal) antibody produced in mouse, clone 2D3, purified immunoglobulin, buffered aqueous solution
SAB3701439
Monoclonal Anti-IL-17A antibody produced in mouse, clone 4H1524.1, purified immunoglobulin
SAB3701458
Monoclonal Anti-IL-17A antibody produced in rat, clone 20B4.G10.F5, purified immunoglobulin
WH0006774M2
Monoclonal Anti-STAT3 antibody produced in mouse, clone 4D6, purified immunoglobulin, buffered aqueous solution
AMAB90777
Monoclonal Anti-STAT3 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL0492, purified immunoglobulin, buffered aqueous glycerol solution
AMAB90776
Monoclonal Anti-STAT3 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL0490, purified immunoglobulin, buffered aqueous glycerol solution