EMAIL THIS PAGE TO A FRIEND
BMC cancer
Relationship between circulating tumor cells and epithelial to mesenchymal transition in early breast cancer.
PMID 26194471
Abstract
Circulating tumor cells (CTCs) play a crucial role in tumor dissemination and are an independent survival predictor in breast cancer (BC) patients. Epithelial to mesenchymal transition (EMT) is involved in cancer invasion and metastasis. The aim of this study was to assess correlation between CTCs and expression of EMT transcription factors TWIST1 and SLUG in breast tumor tissue. This study included 102 early BC patients treated by primary surgery. Peripheral blood mononuclear cells (PBMC) were depleted of hematopoietic cells using RossetteSep™ negative selection kit. RNA extracted from CD45-depleted PBMC was interrogated for expression of EMT (TWIST1, SNAIL1, SLUG, FOXC2 and ZEB1) and epithelial (KRT19) gene transcripts by qRT-PCR. Expression of TWIST1 and SLUG in surgical specimens was evaluated by immunohistochemistry and quantified by multiplicative score. CTCs were detected in 24.5 % patients. CTCs exhibiting only epithelial markers were present in 8.8 % patients, whereas CTCs with only EMT markers were observed in 12.8 % of pts and CTCs co-expressing both markers were detected in 2.9 % pts. We observed lack of correlation between CTCs and expression of TWIST1 and SLUG in breast cancer cells or cancer associated stroma. Lack of correlation was observed for epithelial CTCs as well as for CTCs with EMT. In this translational study, we showed a lack of association between CTCs and expression of EMT-inducing transcription factors, TWIST1 and SLUG, in breast tumor tissue. Despite the fact that EMT is involved in cancer invasion and metastasis our results suggest, that expression of EMT proteins in unselected tumor tissue is not surrogate marker of CTCs with either mesenchymal or epithelial features.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB1305973
ANTI-SLUG (N-TERM K9) antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB1406453
Anti-SNAI2 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB1412527
ANTI-SNAI2 antibody produced in mouse, clone 6A7, purified immunoglobulin, buffered aqueous solution
SAB1412528
ANTI-SNAI2 antibody produced in mouse, clone 4D11, purified immunoglobulin, buffered aqueous solution
AV33302
Anti-SNAI2 antibody produced in rabbit, affinity isolated antibody
SAB1406563
Anti-TWIST1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
T6451
Anti-Twist1 antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
AV37997
Anti-TWIST1 antibody produced in rabbit, IgG fraction of antiserum
SAB2106420
Anti-TWIST1 antibody produced in rabbit, affinity isolated antibody
SAB1411370
Anti-TWIST1 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
WH0006591M5
Monoclonal Anti-SNAI2 antibody produced in mouse, clone 3C12, purified immunoglobulin, buffered aqueous solution
SAB1404377
Monoclonal Anti-SNAI2 antibody produced in mouse, clone 2F11, purified immunoglobulin, buffered aqueous solution
SAB5300372
Monoclonal Anti-SNAI2 antibody produced in mouse, clone 2H5, ascites fluid
WH0007291M1
Monoclonal Anti-TWIST1 antibody produced in mouse, clone 3E11, purified immunoglobulin, buffered aqueous solution
SAB1409772
Monoclonal Anti-TWIST1 antibody produced in mouse, clone 4G10, purified immunoglobulin, buffered aqueous solution
SAB1409773 Monoclonal Anti-TWIST1 antibody produced in mouse, clone 1G2, purified immunoglobulin, buffered aqueous solution
SAB1409774
Monoclonal Anti-TWIST1 antibody produced in mouse, clone 4B4, purified immunoglobulin, buffered aqueous solution
SAB1409775 Monoclonal Anti-TWIST1 antibody produced in mouse, clone 4G11, purified immunoglobulin, buffered aqueous solution
SAB1409776
Monoclonal Anti-TWIST1 antibody produced in mouse, clone 2G12, purified immunoglobulin, buffered aqueous solution
SAB1409777
Monoclonal Anti-TWIST1 antibody produced in mouse, clone 1D3, purified immunoglobulin, buffered aqueous solution
SAB1409778
Monoclonal Anti-TWIST1 antibody produced in mouse, clone 3H5, purified immunoglobulin, buffered aqueous solution
SAB1409779
Monoclonal Anti-TWIST1 antibody produced in mouse, clone 3A2, purified immunoglobulin, buffered aqueous solution
SAB1404499
Monoclonal Anti-TWIST1, (C-terminal) antibody produced in mouse, clone 2F8, purified immunoglobulin, buffered aqueous solution