EMAIL THIS PAGE TO A FRIEND
Molecular oncology
ERBB3 is required for tumor promotion in a mouse model of skin carcinogenesis.
PMID 26194695
Abstract
The epidermal growth factor receptor (EGFR) plays a key role in skin inflammation, wound healing, and carcinogenesis. Less is known about the functions of the structurally related receptor ERBB3 (HER3) in the skin. We assessed the requirement of ERBB3 for skin homeostasis, wound healing, and tumorigenesis by crossing mice carrying a conditional Erbb3 allele with animals expressing cre under the control of the keratin 5 promoter. Erbb3(del) mice, lacking ERBB3 specifically in keratinocytes, showed no obvious abnormalities. The EGFR was upregulated in Erbb3(del) skin, possibly compensating the loss of ERBB3. Nonetheless, healing of full-thickness excisional wounds was negatively affected by ERBB3 deficiency. To analyze the function of ERBB3 during tumorigenesis, we employed the established DMBA/TPA multi-stage chemical carcinogenesis protocol. Erbb3(del) mice remained free of papillomas for a longer time and had significantly reduced tumor burden compared to control littermates. Tumor cell proliferation was considerably reduced in Erbb3(del) mice, and loss of ERBB3 also impaired keratinocyte proliferation after a single application of TPA. In human skin tumor samples, upregulated ERBB3 expression was observed in squamous cell carcinoma, condyloma, and malignant melanoma. Thus, we conclude that ERBB3, while dispensable for the development and the homeostasis of the epidermis and its appendages, is required for proper wound healing and for the progression of skin tumors during multi-stage chemical carcinogenesis in mice. ERBB3 may also be important for human skin cancer progression. The latter effects most probably reflect a key role for ERBB3 in increasing cell proliferation after stimuli as wounding or carcinogenesis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D27802
1,4-Diazabicyclo[2.2.2]octane, ReagentPlus®, ≥99%
C6H12N2
290734
Dabco® 33-LV
C6H12N2