EMAIL THIS PAGE TO A FRIEND
FASEB journal : official publication of the Federation of American Societies for Experimental Biology
Vinculin is required for cell polarization, migration, and extracellular matrix remodeling in 3D collagen.
PMID 26195589
Abstract
Vinculin is filamentous (F)-actin-binding protein enriched in integrin-based adhesions to the extracellular matrix (ECM). Whereas studies in 2-dimensional (2D) tissue culture models have suggested that vinculin negatively regulates cell migration by promoting cytoskeleton-ECM coupling to strengthen and stabilize adhesions, its role in regulating cell migration in more physiologic, 3-dimensional (3D) environments is unclear. To address the role of vinculin in 3D cell migration, we analyzed the morphodynamics, migration, and ECM remodeling of primary murine embryonic fibroblasts (MEFs) with cre/loxP-mediated vinculin gene disruption in 3D collagen I cultures. We found that vinculin promoted 3D cell migration by increasing directional persistence. Vinculin was necessary for persistent cell protrusion, cell elongation, and stable cell orientation in 3D collagen, but was dispensable for lamellipodia formation, suggesting that vinculin-mediated cell adhesion to the ECM is needed to convert actin-based cell protrusion into persistent cell shape change and migration. Consistent with this finding, vinculin was necessary for efficient traction force generation in 3D collagen without affecting myosin II activity and promoted 3D collagen fiber alignment and macroscopical gel contraction. Our results suggest that vinculin promotes directionally persistent cell migration and tension-dependent ECM remodeling in complex 3D environments by increasing cell-ECM adhesion and traction force generation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB1305939
ANTI-MYL12B (N-TERM) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
HPA019763
Anti-MYL2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA039262
Anti-MYL2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1403431
Monoclonal Anti-MRLC2, (C-terminal) antibody produced in mouse, clone 1B22, purified immunoglobulin, buffered aqueous solution
WH0004633M1
Monoclonal Anti-MYL2 antibody produced in mouse, clone 3B9-B4, purified immunoglobulin, buffered aqueous solution
SAB5300243
Monoclonal Anti-MYL2 antibody produced in mouse, clone 7C9, ascites fluid
SAB1401660 Monoclonal Anti-MYLPF antibody produced in mouse, clone 3H3, purified immunoglobulin, buffered aqueous solution
GS203NPEAQU
Whatman® Mini-UniPrep® G2 slit septum, PVDF membrane, pore size 0.2 μm, clear vial, pack of 100
GS503NPEAQU
Whatman® Mini-UniPrep® G2 slit septum, PVDF membrane, pore size 0.2 μm, clear vial, pack of 1000
GS203NPUAQU
Whatman® Mini-UniPrep® G2 slit septum, PVDF membrane, pore size 0.45 μm, clear vial, pack of 100
GS503NPUAQU
Whatman® Mini-UniPrep® G2 slit septum, PVDF membrane, pore size 0.45 μm, clear vial, pack of 1000
GS203NPEAQUSP
Whatman® Mini-UniPrep® G2 slit septum starter pack, supplied with single vial compressor, PVDF membrane, pore size 0.20 μm, clear vial, pack of 100
GS203NPUAQUSP
Whatman® Mini-UniPrep® G2 slit septum starter pack, supplied with single vial compressor, PVDF membrane, pore size 0.45 μm, clear vial, pack of 100
WHAGN203NPEAQU
Whatman® Mini-UniPrep® G2 standard septum, PVDF membrane, pore size 0.2 μm, clear vial color, pack of 100
WHAGN203NPUAQU
Whatman® Mini-UniPrep® G2 standard septum, PVDF membrane, pore size 0.45 μm, clear vial color, pack of 100
GN503NPEAQU
Whatman® Mini-UniPrep® G2 standard septum, PVDF membrane, pore size 0.2 μm, clear vial, pack of 1000
GN203APEAQU
Whatman® Mini-UniPrep® G2 standard septum, PVDF membrane, pore size 0.2 μm, amber vial, pack of 100
GN503NPUAQU
Whatman® Mini-UniPrep® G2 standard septum, PVDF membrane, pore size 0.45 μm, clear vial, pack of 1000
WHAGN203APEAQUSP
Whatman® Mini-UniPrep® G2 standard septum starter pack, supplied with single vial compressor, PVDF membrane, pore size 0.2 μm, amber vial color, pack of 100
Z759406
Whatman® Mini-UniPrep® G2 standard septum starter pack, supplied with single vial compressor, PVDF membrane, pore size 0.2 μm, clear vial color, pack of 100
WHAGN203NPEAQUSP
Whatman® Mini-UniPrep® G2 standard septum starter pack, supplied with single vial compressor, PVDF membrane, pore size 0.2 μm, clear vial color, pack of 100
WHAGN203NPUAQUSP
Whatman® Mini-UniPrep® G2 standard septum starter pack, supplied with single vial compressor, PVDF membrane, pore size 0.45 μm, clear vial color, pack of 100