EMAIL THIS PAGE TO A FRIEND
Natural product communications
Chemical Constituents of the Underground Parts of Iris florentina and their Cytotoxic Activity.
PMID 26197526
Abstract
Three new isoflavonoid glycosides (1, 5, and 9) and 10 known compounds (2-4, 6-8, and 10-13) were isolated from the underground parts of Iris florentina (Iridaceae). The structures of the new compounds were determined based on extensive spectroscopic data and the results of hydrolytic cleavage. The isolated compounds and the aglycones were evaluated for cytotoxic activity against HL-60 human promyelocytic leukemia cells. Compound 12 induced apoptotic cell death in the HL-60 cells.