EMAIL THIS PAGE TO A FRIEND
EMBO molecular medicine
Folate levels modulate oncogene-induced replication stress and tumorigenicity.
PMID 26197802
Abstract
Chromosomal instability in early cancer stages is caused by replication stress. One mechanism by which oncogene expression induces replication stress is to drive cell proliferation with insufficient nucleotide levels. Cancer development is driven by alterations in both genetic and environmental factors. Here, we investigated whether replication stress can be modulated by both genetic and non-genetic factors and whether the extent of replication stress affects the probability of neoplastic transformation. To do so, we studied the effect of folate, a micronutrient that is essential for nucleotide biosynthesis, on oncogene-induced tumorigenicity. We show that folate deficiency by itself leads to replication stress in a concentration-dependent manner. Folate deficiency significantly enhances oncogene-induced replication stress, leading to increased DNA damage and tumorigenicity in vitro. Importantly, oncogene-expressing cells, when grown under folate deficiency, exhibit a significantly increased frequency of tumor development in mice. These findings suggest that replication stress is a quantitative trait affected by both genetic and non-genetic factors and that the extent of replication stress plays an important role in cancer development.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C6891
5-Chloro-2′-deoxyuridine, thymidine analog
C9H11ClN2O5
B4436
Anti-53BP1 (C-terminal) antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
B4561
Anti-53BP1 (N-terminal) antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
GW21178 Anti-ATM antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB3500147
Anti-ATM antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB4501968
Anti-ATM antibody produced in rabbit, affinity isolated antibody
A6218 Anti-ATM antibody, Mouse monoclonal, clone SYML6A10, purified from hybridoma cell culture
A6093 Anti-ATM antibody, Mouse monoclonal, clone SYR6D4, purified from hybridoma cell culture
SAB1400043
Anti-CCNE1 antibody produced in mouse, IgG fraction of antiserum, buffered aqueous solution
HPA018169
Anti-CCNE1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1400044
Anti-CCNE1 antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB4300390
Anti-CHEK1 (Ab-317) antibody produced in rabbit, affinity isolated antibody
AV32589
Anti-CHEK1 antibody produced in rabbit, affinity isolated antibody
SAB2104134
Anti-CHEK1 antibody produced in rabbit, affinity isolated antibody
SAB4500207
Anti-CHK1 antibody produced in rabbit, affinity isolated antibody
SAB4500208
Anti-CHK1 antibody produced in rabbit, affinity isolated antibody
SAB4500209
Anti-CHK1 antibody produced in rabbit, affinity isolated antibody
SAB4500210
Anti-CHK1 antibody produced in rabbit, affinity isolated antibody
SAB4500211
Anti-CHK1 antibody produced in rabbit, affinity isolated antibody
SAB4503515
Anti-Cyclin E1 antibody produced in rabbit, affinity isolated antibody
SAB4503516
Anti-Cyclin E1 antibody produced in rabbit, affinity isolated antibody
SAB4503514
Anti-Cyclin E1, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4300514
Anti-H2AFX (Ab-139) antibody produced in rabbit, affinity isolated antibody
SAB4501369
Anti-Histone H2AX , C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB1301801
ANTI-POTEF (CENTER) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB1406364
Anti-RAD51 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB2101936
Anti-RAD51 antibody produced in rabbit, affinity isolated antibody
SAB2700175
Anti-RAD51 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
HPA039310
Anti-RAD51 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4502565
Anti-RAD51, C-Terminal antibody produced in rabbit, affinity isolated antibody