EMAIL THIS PAGE TO A FRIEND
Marine drugs
Bioactive 7-Oxabicyclic[6.3.0]lactam and 12-Membered Macrolides from a Gorgonian-Derived Cladosporium sp. Fungus.
PMID 26198234
Abstract
One new bicyclic lactam, cladosporilactam A (1), and six known 12-membered macrolides (2-7) were isolated from a gorgonian-derived Cladosporium sp. fungus collected from the South China Sea. Their complete structural assignments were elucidated by comprehensive spectroscopic investigation. Quantum chemistry calculations were used in support of the structural determination of 1. The absolute configuration of 1 was determined by calculation of its optical rotation. Cladosporilactam A (1) was the first example of 7-oxabicyclic[6.3.0]lactam obtained from a natural source. Compound 1 exhibited promising cytotoxic activity against cervical cancer HeLa cell line with an IC50 value of 0.76 μM.