EMAIL THIS PAGE TO A FRIEND
Nature communications
DOT1L cooperates with the c-Myc-p300 complex to epigenetically derepress CDH1 transcription factors in breast cancer progression.
PMID 26199140
Abstract
DOT1L has emerged as an anticancer target for MLL-associated leukaemias; however, its functional role in solid tumours is largely unknown. Here we identify that DOT1L cooperates with c-Myc and p300 acetyltransferase to epigenetically activate epithelial-mesenchymal transition (EMT) regulators in breast cancer progression. DOT1L recognizes SNAIL, ZEB1 and ZEB2 promoters via interacting with the c-Myc-p300 complex and facilitates lysine-79 methylation and acetylation towards histone H3, leading to the dissociation of HDAC1 and DNMT1 in the regions. The upregulation of these EMT regulators by the DOT1L-c-Myc-p300 complex enhances EMT-induced breast cancer stem cell (CSC)-like properties. Furthermore, in vivo orthotopic xenograft models show that DOT1L is required for malignant transformation of breast epithelial cells and breast tumour initiation and metastasis. Clinically, DOT1L expression is associated with poorer survival and aggressiveness of breast cancers. Collectively, we suggest that cooperative effect of DOT1L and c-Myc-p300 is critical for acquisition of aggressive phenotype of breast cancer by promoting EMT/CSC.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
660124
4,4′-Bis(N-carbazolyl)-1,1′-biphenyl, 97%
C36H24N2
699195
4,4′-Bis(N-carbazolyl)-1,1′-biphenyl, sublimed grade, 99.9% trace metals basis
C36H24N2
06-599
Anti-acetyl-Histone H3 Antibody, 100 µg, Detect acetyl-Histone H3 with Anti-acetyl-Histone H3 Antibody (Rabbit Polyclonal Antibody), that has been shown to work in WB, ICC, ChIP, ChIP-seq.
MAB1501R
Anti-Actin Antibody, near a.a. 50-70, clone C4, 100 µg, This Anti-Actin Antibody, near a.a. 50-70, clone C4 is validated for use in ELISA, IC, IH, IH(P), WB for the detection of Actin and has published in over 70 citations.
05-257
Anti-p300 CT Antibody, clone RW128, 200 µg, Use Anti-p300 CT Antibody, clone RW128 (Mouse Monoclonal Antibody) validated in ChIP, IP, WB to detect p300 CT also known as KAT3B.
GW21186 Anti-CDH1 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB2501665
Anti-CDH1 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB2701861
Anti-cdh1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
HPA004812
Anti-CDH1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2104222
Anti-CDH1 antibody produced in rabbit, affinity isolated antibody
SAB4300693
Anti-CDH1 antibody produced in rabbit, affinity isolated antibody
SAB2107370
Anti-CDH1 antibody produced in rabbit, affinity isolated antibody
SAB4501941
Anti-MYC, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4503661
Anti-MYC, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4503662
Anti-MYC, C-Terminal antibody produced in rabbit, affinity isolated antibody
P9795 Anti-Rabbit IgG (whole molecule)−R-Phycoerythrin antibody produced in goat, affinity isolated antibody, buffered aqueous solution
CKOZFN1041 CompoZr® Knockout ZFN Kit, human MYC (NM_002467)
D1515
Doxorubicin hydrochloride, 98.0-102.0% (HPLC)
C27H29NO11 · HCl
44583
Doxorubicin hydrochloride, suitable for fluorescence, 98.0-102.0% (HPLC)
C27H29NO11 · HCl
E1024
Estradiol, meets USP testing specifications
C18H24O2
EHU092391 MISSION® esiRNA, esiRNA human DOT1L (esiRNA1)
C7855
Monoclonal Anti-Cdh1 antibody produced in mouse, ~2 mg/mL, clone DCS-266, purified immunoglobulin, buffered aqueous solution
WH0000999M1
Monoclonal Anti-CDH1 antibody produced in mouse, clone 3F4, purified immunoglobulin, buffered aqueous solution
AMAB90862
Monoclonal Anti-CDH1 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL1170, purified immunoglobulin, buffered aqueous glycerol solution
AMAB90863
Monoclonal Anti-CDH1 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL1172, purified immunoglobulin, buffered aqueous glycerol solution
AMAB90865
Monoclonal Anti-CDH1 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL1180, purified immunoglobulin, buffered aqueous glycerol solution
SAB4700237 Monoclonal Anti-CDH1 antibody produced in mouse, clone 67A4, purified immunoglobulin, buffered aqueous solution
WH0084444M1
Monoclonal Anti-DOT1L antibody produced in mouse, clone 6A6, purified immunoglobulin, buffered aqueous solution
SAB1405309
Monoclonal Anti-DOT1L antibody produced in mouse, clone 8B2, purified immunoglobulin, buffered aqueous solution
SAB4700447 Monoclonal Anti-MYC , (C-terminal) antibody produced in mouse, clone 9E10, purified immunoglobulin, buffered aqueous solution