EMAIL THIS PAGE TO A FRIEND
Beilstein journal of organic chemistry
Regioselective synthesis of chiral dimethyl-bis(ethylenedithio)tetrathiafulvalene sulfones.
PMID 26199666
Abstract
Enantiopure (R,R) and (S,S)-dimethyl-bis(ethylenedithio)tetrathiafulvalene monosulfones have been synthesized by the aerial oxidation of the chiral dithiolates generated from the propionitrile-protected precursors. Both enantiomers crystallize in the orthorhombic chiral space group P212121. They show a boat-type conformation of the TTF moiety, a rather rigid dithiin sulfone ring and the methyl groups in a bisequatorial conformation. Cyclic voltammetry measurements indicate fully reversible oxidation in radical cation and dication species.