EMAIL THIS PAGE TO A FRIEND
The EMBO journal
Triggered Ca2+ influx is required for extended synaptotagmin 1-induced ER-plasma membrane tethering.
PMID 26202220
Abstract
The extended synaptotagmins (E-Syts) are ER proteins that act as Ca(2+)-regulated tethers between the ER and the plasma membrane (PM) and have a putative role in lipid transport between the two membranes. Ca(2+) regulation of their tethering function, as well as the interplay of their different domains in such function, remains poorly understood. By exposing semi-intact cells to buffers of variable Ca(2+) concentrations, we found that binding of E-Syt1 to the PI(4,5)P2-rich PM critically requires its C2C and C2E domains and that the EC50 of such binding is in the low micromolar Ca(2+) range. Accordingly, E-Syt1 accumulation at ER-PM contact sites occurred only upon experimental manipulations known to achieve these levels of Ca(2+) via its influx from the extracellular medium, such as store-operated Ca(2+) entry in fibroblasts and membrane depolarization in β-cells. We also show that in spite of their very different physiological functions, membrane tethering by E-Syt1 (ER to PM) and by synaptotagmin (secretory vesicles to PM) undergo a similar regulation by plasma membrane lipids and cytosolic Ca(2+).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
HPA016858
Anti-ESYT1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA002132
Anti-ESYT2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2500381
Anti-FAM62A antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB1407201
Anti-FAM62A antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB1409571
Anti-FAM62B antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB2104881
Anti-FAM62B antibody produced in rabbit, affinity isolated antibody
S6197
Anti-STIM1 (C-terminal) antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
S6072
Anti-STIM1 (N-terminal) antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
SAB2501933 Anti-STIM1 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
HPA011088
Anti-STIM1 antibody produced in rabbit, Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA012123
Anti-STIM1 antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB3500365
Anti-STIM1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
L5163
Latrunculin A, from sea sponge, ≥85% (HPLC), waxy solid
C22H31NO5S
WH0057488M2
Monoclonal Anti-FAM62B antibody produced in mouse, clone 4C3, purified immunoglobulin, buffered aqueous solution
SAB1405210
Monoclonal Anti-FAM62B, (N-terminal) antibody produced in mouse, clone 1E10, purified immunoglobulin, buffered aqueous solution
WH0006786M1
Monoclonal Anti-STIM1 antibody produced in mouse, clone 5A2, purified immunoglobulin, buffered aqueous solution
SAB1404421
Monoclonal Anti-STIM1, (C-terminal) antibody produced in mouse, clone 6C6, purified immunoglobulin, buffered aqueous solution
C8031
N6-Cyclopentyladenosine, solid
C15H21N5O4
N0128
Nitrilotriacetic acid disodium salt, Sigma Grade, ≥99%
C6H7NNa2O6
N0253
Nitrilotriacetic acid trisodium salt, Sigma Grade, ≥98%
C6H6NNa3O6