EMAIL THIS PAGE TO A FRIEND
Thrombosis and haemostasis
HMGB1 binds to activated platelets via the receptor for advanced glycation end products and is present in platelet rich human coronary artery thrombi.
PMID 26202300
Abstract
High mobility group box 1 (HMGB1) acts as both a nuclear protein that regulates gene expression, as well as a pro-inflammatory alarmin that is released from necrotic or activated cells. Recently, HMGB1-expression in human atherosclerotic plaques was identified. Therapeutic blockade of HMGB1 reduced the development of diet-induced atherosclerosis in ApoE knockout mice. Thus, we hypothesised an interaction between HMGB1 and activated platelets. Binding of recombinant HMGB1 to platelets was assessed by flow cytometry. HMGB1 bound to thrombin-activated human platelets (MFI 2.49 vs 25.01, p=0.0079). Blood from wild-type, TLR4 and RAGE knockout mice was used to determine potential HMGB1 receptors on platelets. HMGB1 bound to platelets from wild type C57Bl6 (MFI 2.64 vs 20.3, p< 0.05), and TLR4-/- mice (MFI 2.11 vs 25.65, p< 0.05) but failed to show binding to platelets from RAGE-/- mice (p > 0.05). RAGE expression on human platelets was detected by RT-PCR with mRNA extracted from highly purified platelets and confirmed by Western blot and immunofluorescence microscopy. Platelet activation increased RAGE surface expression (MFI 4.85 vs 6.74, p< 0.05). Expression of HMGB1 in human coronary artery thrombi was demonstrated by immunohistochemistry and revealed high expression levels. Platelets bind HMGB1 upon thrombin-induced activation. Platelet specific expression of RAGE could be detected at the mRNA and protein level and is involved in the binding of HMGB1. Furthermore, platelet activation up-regulates platelet surface expression of RAGE. HMGB1 is highly expressed in platelet-rich human coronary artery thrombi pointing towards a central role for HMGB1 in atherothrombosis, thereby suggesting the possibility of platelet targeted anti-inflammatory therapies for atherothrombosis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB3700280 Anti-goat IgG (H+L) antibody produced in donkey, affinity isolated antibody, buffered aqueous solution
SAB3700328 Anti-Goat IgG (H+L), F(ab) fragment- antibody produced in donkey, affinity isolated antibody, buffered aqueous solution
AV38110
Anti-HMGB1 (AB2) antibody produced in rabbit, IgG fraction of antiserum
SAB2701685
Anti-hmgb1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2701809
Anti-hmgb1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2101049
Anti-HMGB1 antibody produced in rabbit, affinity isolated antibody
SAB4501401
Anti-HMGB1 antibody produced in rabbit, affinity isolated antibody
HPA013796
Anti-P2RY12 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA014518
Anti-P2RY12 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
P6997 Anti-P2Y12 Platelet ADP Receptor antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB1406424
Anti-SELP antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA002655
Anti-SELP antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA005990
Anti-SELP antibody produced in rabbit, Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
H4652 HMG-1 human, lyophilized powder, ≥90% (SDS-PAGE), Histidine-tagged, recombinant, expressed in E. coli
H9537
Monoclonal Anti-HMG-1 antibody produced in mouse, clone HAP46.5, purified from hybridoma cell culture
WH0003146M8
Monoclonal Anti-HMGB1 antibody produced in mouse, clone 2F6, purified immunoglobulin, buffered aqueous solution
SAB1403925
Monoclonal Anti-HMGB1 antibody produced in mouse, clone 1D5, purified immunoglobulin, buffered aqueous solution
P0115
PAC-1, ≥98% (HPLC)
C23H28N4O2