EMAIL THIS PAGE TO A FRIEND
Scientific reports
IRGM1 enhances B16 melanoma cell metastasis through PI3K-Rac1 mediated epithelial mesenchymal transition.
PMID 26202910
Abstract
Melanoma is one of the most aggressive skin cancers and is well known for its high metastatic rate. Studies have shown that epithelial mesenchymal transition (EMT) is essential for melanoma cell metastasis. However, the molecular mechanisms underlying EMT are still not fully understood. We have shown that IRGM1, a member of immunity-related GTPase family that regulates immune cell motility, is highly expressed by melanoma cells. The current study aimed to explore whether and how IRGM1 may regulate melanoma cell metastasis. To test this, we modified IRGM1 expression in B16 melanoma cells. We found that over-expression of IRGM1 substantially enhanced pulmonary metastasis in vivo. In keeping with that, knocking-in IRGM1 strongly enhanced while knocking-down IRGM1 impaired B16 cell migration and invasion ability in vitro. Interestingly, we observed that IRGM1 enhanced F-actin polymerization and triggers epithelial mesenchymal transition (EMT) through a mechanism involved in PIK3CA mediated Rac1 activation. Together, these data reveals a novel molecular mechanism that involved in melanoma metastasis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
PRS4543
Anti-IRGM (ab1) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
PRS4545
Anti-IRGM (ab2) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2900758
Anti-IRGM antibody produced in rabbit, affinity isolated antibody
SAB1307004
ANTI-IRGM(C-TERMINAL) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
GW21071 Anti-PI3K antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
HPA009985
Anti-PIK3CA antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2701957
Anti-PIK3CA antibody produced in rabbit
SAB4300461
Anti-RAC1 (Ab-71) antibody produced in rabbit, affinity isolated antibody
GW22212F Anti-RAC1 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB2106168
Anti-RAC1 antibody produced in rabbit, affinity isolated antibody
SAB4502560
Anti-RAC1 antibody produced in rabbit, affinity isolated antibody
WH0005290M1 Monoclonal Anti-PIK3CA antibody produced in mouse, clone 3G3, purified immunoglobulin, buffered aqueous solution
SAB5300225
Monoclonal Anti-PIK3CA antibody produced in mouse, clone 4F3, purified immunoglobulin, buffered aqueous solution
N2913 N-TER Nanoparticle siRNA Transfection System