EMAIL THIS PAGE TO A FRIEND
Journal of environmental management
Novel Chryseobacterium sp. PYR2 degrades various organochlorine pesticides (OCPs) and achieves enhancing removal and complete degradation of DDT in highly contaminated soil.
PMID 26203874
Abstract
Long term residues of organochlorine pesticides (OCPs) in soils are of great concerning because they seriously threaten food security and human health. This article focuses on isolation of OCP-degrading strains and their performance in bioremediation of contaminated soil under ex situ conditions. A bacterium, Chryseobacterium sp. PYR2, capable of degrading various OCPs and utilizing them as a sole carbon and energy source for growth, was isolated from OCP-contaminated soil. In culture experiments, PYR2 degraded 80-98% of hexachlorocyclohexane (HCH) or 1,1,1-trichloro-2,2-bis (4-chlorophenyl) ethane (DDT) isomers (50xa0mgxa0L(-1)) in 30 days. A pilot-scale ex situ bioremediation study of highly OCP-contaminated soil augmented with PYR2 was performed. During the 45-day experimental period, DDT concentration was reduced by 80.3% in PYR2-augmented soils (35.37xa0mgxa0kg(-1) to 6.97xa0mgxa0kg(-1)) but by only 57.6% in control soils. Seven DDT degradation intermediates (metabolites) were detected and identified in PYR2-augmented soils: five by GC/MS: 1,1-dichloro-2,2-bis (4-chlorophenyl) ethane (DDD), 1,1-dichloro-2,2-bis (4-chlorophenyl) ethylene (DDE), 1-chloro-2,2-bis (4-chlorophenyl) ethylene (DDMU), 1-chloro-2,2-bis (4-chlorophenyl) ethane (DDMS), and dichlorobenzophenone (DBP); and two by LC/MS: 4-chlorobenzoic acid (PCBA) and 4-chlorophenylacetic acid (PCPA). Levels of metabolites were fairly stable in control soils but varied greatly with time in PYR2-augmented soils. Levels of DDD, DDMU, and DDE in PYR2-augmented soils increased from day 0 to day 30 and then decreased by day 45. A DDT biodegradation pathway is proposed based on our identification of DDT metabolites in PYR2-augmented systems. PYR2 will be useful in future studies of OCP biodegradation and in bioremediation of OCP-contaminated soils.