EMAIL THIS PAGE TO A FRIEND
Biomaterials
Oligonucleotide aptamer-drug conjugates for targeted therapy of acute myeloid leukemia.
PMID 26204224
Abstract
Oligonucleotide aptamers can specifically bind biomarkers on cancer cells and can be readily chemically modified with different functional molecules for personalized medicine. To target acute myeloid leukemia (AML) cells, we developed a single-strand DNA aptamer specific for the biomarker CD117, which is highly expressed on AML cells. Sequence alignment revealed that the aptamer contained a G-rich core region with a well-conserved functional G-quadruplex structure. Functional assays demonstrated that this synthetic aptamer was able to specifically precipitate CD117 proteins from cell lysates, selectively bound cultured and patient primary AML cells with high affinity (Kdxa0
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
21888
5(6)-Carboxyfluorescein diacetate N-succinimidyl ester, BioReagent, suitable for fluorescence, ≥90% (HPLC)
C29H19NO11
87444
5-Carboxy-fluorescein diacetate N-succinimidyl ester, for fluorescence, ≥95.0% (HPLC)
C29H19NO11
SAB4300489
Anti-KIT (Ab-721) antibody produced in rabbit, affinity isolated antibody
SAB4300593
Anti-KIT (Ab-936) antibody produced in rabbit, affinity isolated antibody
GW21114 Anti-KIT antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB1412042
ANTI-KIT antibody produced in mouse, clone 1G1, purified immunoglobulin, buffered aqueous solution
SAB1412043
ANTI-KIT antibody produced in mouse, clone 5F6, purified immunoglobulin, buffered aqueous solution
SAB1412044
ANTI-KIT antibody produced in mouse, clone 5A11, purified immunoglobulin, buffered aqueous solution
SAB1412045
ANTI-KIT antibody produced in mouse, clone 2C3, purified immunoglobulin, buffered aqueous solution
SAB1412046
ANTI-KIT antibody produced in mouse, clone 1D3, purified immunoglobulin, buffered aqueous solution
SAB1412140
ANTI-KIT antibody produced in mouse, clone 4F7, purified immunoglobulin, buffered aqueous solution
SAB1412141
ANTI-KIT antibody produced in mouse, clone 3A8, purified immunoglobulin, buffered aqueous solution
HPA004471
Anti-KIT antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4501647
Anti-KIT, C-Terminal antibody produced in rabbit, affinity isolated antibody
M7824
Methotrexate, PharmaGrade, Manufactured under appropriate controls for use as a raw material in pharma or biopharmaceutical production, meets EP, USP testing specifications
C20H22N8O5
M4010
Methotrexate, meets EP, USP testing specifications
C20H22N8O5 · xH2O
M9929
Methotrexate, meets USP testing specifications
C20H22N8O5 · xH2O
SAB4700750
Monoclonal Anti-CD117 antibody produced in mouse, clone 104D2, purified immunoglobulin, buffered aqueous solution
SAB4700284 Monoclonal Anti-CD15 antibody produced in mouse, clone MEM-158, purified immunoglobulin, buffered aqueous solution
WH0003815M2
Monoclonal Anti-KIT antibody produced in mouse, clone 6F2, purified immunoglobulin, buffered aqueous solution
AMAB90901
Monoclonal Anti-KIT antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL1657, purified immunoglobulin, buffered aqueous glycerol solution
AMAB90904
Monoclonal Anti-KIT antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL1667, purified immunoglobulin, buffered aqueous glycerol solution
SAB1404007
Monoclonal Anti-KIT antibody produced in mouse, clone 6G12, purified immunoglobulin, buffered aqueous solution
SAB5300439
Monoclonal Anti-KIT antibody produced in mouse, clone 8D7, ascites fluid
SAB5300440
Monoclonal Anti-KIT antibody produced in mouse, clone 1C5, ascites fluid
T3516
Thioflavin T, used as stain for amyloid
C17H19ClN2S