PloS one
Genome-Wide Mapping of Collier In Vivo Binding Sites Highlights Its Hierarchical Position in Different Transcription Regulatory Networks.
PMID 26204530
Abstract
Collier, the single Drosophila COE (Collier/EBF/Olf-1) transcription factor, is required in several developmental processes, including head patterning and specification of muscle and neuron identity during embryogenesis. To identify direct Collier (Col) targets in different cell types, we used ChIP-seq to map Col binding sites throughout the genome, at mid-embryogenesis. In vivo Col binding peaks were associated to 415 potential direct target genes. Gene Ontology analysis revealed a strong enrichment in proteins with DNA binding and/or transcription-regulatory properties. Characterization of a selection of candidates, using transgenic CRM-reporter assays, identified direct Col targets in dorso-lateral somatic muscles and specific neuron types in the central nervous system. These data brought new evidence that Col direct control of the expression of the transcription regulators apterous and eyes-absent (eya) is critical to specifying neuronal identities. They also showed that cross-regulation between col and eya in muscle progenitor cells is required for specification of muscle identity, revealing a new parallel between the myogenic regulatory networks operating in Drosophila and vertebrates. Col regulation of eya, both in specific muscle and neuronal lineages, may illustrate one mechanism behind the evolutionary diversification of Col biological roles.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB1401144
Anti-FPR2 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
HPA029154 Anti-FPR2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
AV43141
Anti-MKRN1 (AB1) antibody produced in rabbit, IgG fraction of antiserum
AV43142
Anti-MKRN1 (AB2) antibody produced in rabbit, IgG fraction of antiserum
SAB2501704
Anti-MKRN1 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB2501717
Anti-MKRN1 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB2102989
Anti-TRIM27 antibody produced in rabbit, affinity isolated antibody
SAB2700796
Anti-TRIM27 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2700798
Anti-TRIM27 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2700971
Anti-TRIM27 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
HPA048684
Anti-TRIM27 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1403837
Monoclonal Anti-FPRL1 antibody produced in mouse, clone 2G8, purified immunoglobulin, buffered aqueous solution
SAB1402202
Monoclonal Anti-FPRL1 antibody produced in mouse, clone 2D8, purified immunoglobulin, buffered aqueous solution
WH0023608M1
Monoclonal Anti-MKRN1 antibody produced in mouse, clone 4E9, purified immunoglobulin, buffered aqueous solution