EMAIL THIS PAGE TO A FRIEND
Archiv der Pharmazie
Synthesis and Evaluation of a Series of Oleanolic Acid Saponins as α-Glucosidase and α-Amylase Inhibitors.
PMID 26207761
Abstract
Sixteen naturally occurring oleanolic acid saponins and their derivatives were synthesized in an efficient and practical strategy, and their inhibitory activities against α-glucosidase and α-amylase were evaluated in vitro. Among all the compounds, 28-O-monoglucoside 8 exhibited remarkably potent inhibitory activity against α-glucosidase with an IC50 value of 87.3 µM, which was fivefold stronger than that of the antidiabetic acarbose. Based on the preliminary structure-activity relationships, for 28-O-monoglucosides, the presence of a terminal α-l-rhamnopyranosyl residue enhanced the α-glucosidase and α-amylase inhibitory activities. Furthermore, for 3,28-O-bidesmosides, sugar-substituted moieties attached to the C-3 and C-28 positions of the oleanolic acid scaffold are helpful to increase the inhibitory activities against α-amylase and α-glucosidase.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
O5504
Oleanolic acid, ≥97%
C30H48O3