EMAIL THIS PAGE TO A FRIEND
Cellular & molecular biology letters
Neurotrophine-3 may contribute to neuronal differentiation of mesenchymal stem cells through the activation of the bone morphogenetic protein pathway.
PMID 26208387
Abstract
We investigated whether neurotrophin-3 (NT-3) can promote differentiation of mouse bone mesenchymal stem cells (MSCs) into neurons via the bone morphogenetic protein pathway. MSCs were prepared from rat bone marrow and either transfected with pIRES2-EGFP or pIRES2-EGFP-NT-3 or treated with bone morphogenetic protein 4. The pIRES2-EGFP-NT-3-transfected MSCs further underwent noggin treatment or siRNA-mediated knockout of the TrkC gene or were left untreated. Immunofluorescence staining, real-time PCR and Western blot analyses were performed to evaluate the transcription and expression of neural-specific genes and BMP-Smad signaling. MSCs were efficiently transduced by the NT-3 gene via pIRES2-EGFP vectors. pIRES2- EGFP-NT-3 could initiate the transcription and expression of neural-specific genes, including nestin, NSE and MAP-2, and stimulate BMP-Smad signaling. The transcription and expression of neural-specific genes and BMP-Smad signaling were significantly suppressed by siRNA-mediated knockdown of the TrkC gene of MSCs. These findings suggest that the BMP signaling pathway may be a key regulatory point in NT-3-transfected neuronal differentiation of MSCs. The BMP and neurotrophin pathways contribute to a tightly regulated signaling network that directs the precise connections between neuronal differentiation of MSCs and their targets.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
345571
1,5,7-Triazabicyclo[4.4.0]dec-5-ene, 98%
C7H13N3
SAB4200516
Anti-BMP4 (N-terminal) antibody produced in rabbit, ~1.0 mg/mL, affinity isolated antibody
SAB4200556
Anti-BMP4 antibody produced in rabbit, ~1.0 mg/mL, affinity isolated antibody
SAB2700755
Anti-BMP4 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2700756
Anti-BMP4 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB1300906
Anti-NTF3 (C-term) antibody produced in rabbit, saturated ammonium sulfate (SAS) precipitated, buffered aqueous solution
SAB1300907
Anti-NTF3 (center) antibody produced in rabbit, saturated ammonium sulfate (SAS) precipitated, buffered aqueous solution
SAB1400188
Anti-NTF3 antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
HPA032000
Anti-NTF3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2701787
Anti-rpl3 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
HPA003365
Anti-RPL3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
AV48157
Anti-RPL3 antibody produced in rabbit, affinity isolated antibody
SAB2102052
Anti-RPL3 antibody produced in rabbit, affinity isolated antibody
SAB4502653
Anti-RPL3 antibody produced in rabbit, affinity isolated antibody
SAB4300561
Anti-SMAD1 (Ab-465) antibody produced in rabbit, affinity isolated antibody
AV38693
Anti-SMAD1 (AB1) antibody produced in rabbit, IgG fraction of antiserum
AV38692
Anti-SMAD1 (AB2) antibody produced in rabbit, IgG fraction of antiserum
SAB4200377
Anti-SMAD1 (Internal region) antibody produced in rabbit, ~1.0 mg/mL, affinity isolated antibody
SAB1412181
ANTI-SMAD1 antibody produced in mouse, clone 1C7, purified immunoglobulin, buffered aqueous solution
SAB1412594
ANTI-SMAD1 antibody produced in mouse, clone 3G4, purified immunoglobulin, buffered aqueous solution
SAB1412595
ANTI-SMAD1 antibody produced in mouse, clone 4C12, purified immunoglobulin, buffered aqueous solution
SAB1412656
ANTI-SMAD1 antibody produced in mouse, clone 1B8, purified immunoglobulin, buffered aqueous solution
AV32003
Anti-SMAD1 antibody produced in rabbit, affinity isolated antibody
SAB4200378
Anti-SMAD1 antibody produced in rabbit, ~1.0 mg/mL, affinity isolated antibody
SAB4501802
Anti-SMAD1 antibody produced in rabbit, affinity isolated antibody
SAB4501801
Anti-SMAD1, C-Terminal antibody produced in rabbit, affinity isolated antibody
EHU098101 MISSION® esiRNA, esiRNA human NTRK3 (esiRNA2)
EHU092181 MISSION® esiRNA, esiRNA human NTRK3 (esiRNA1)
EMU040761 MISSION® esiRNA, esiRNA targeting mouse Ntrk3 (esiRNA1)
SAB3300014
Monoclonal Anti-BMP4 antibody produced in mouse, clone 6B7, purified immunoglobulin, buffered aqueous glycerol solution