EMAIL THIS PAGE TO A FRIEND
Journal of molecular histology
TGFβ modulates inflammatory cytokines and growth factors to create premetastatic microenvironment and stimulate lung metastasis.
PMID 26208571
Abstract
The formation of tumor-promoting premetastatic microenvironment plays a pivotal role on metastatic progression. Understanding how the primary tumor can promote the formation of premetastatic microenvironment in the lung will aid discovery of a final cure for metastatic breast cancer. The murine 4T1 mammary carcinoma cells were injected into the mammary fat pads of the BALB/c mice. Days 0-14 were considered the premetastatic phase. Lung tissues were examined using hematoxylin-eosin staining and transmission electron microscopy. After intravenous injection of TGFβ1 pretreated 4T1 cells, the relative pulmonary vascular permeability was quantified, the extravasation, survival, and proliferation of tumor cells in premetastatic lungs were evaluated, and the levels of S100A8, S100A9, VEGF, and Angpt2 were detected in tumor-bearing mice. The results showed that during the premetastatic phase, an inflammatory response and inflammation-induced vascular hyperpermeability were established, leading to an abnormal pulmonary microenvironment, which facilitated extravasation of circulating tumor cells, and subsequent survival and proliferation of metastatic tumor cells in a TGFβ-dependent manner. Moreover, the expressions of S100A8, S100A9, VEGF, and Angpt2 were increased, and an induction of these genes by TGFβ was further observed in premetastatic lungs. Thus, this study demonstrated that TGFβ promoted the creation of premetastatic microenvironment by modulating certain crucial inflammatory cytokines and growth factors, and finally enhanced the ability of circulating cells to seed the lung.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A0851 Anti-Angiopoietin-2 antibody produced in goat, affinity isolated antibody, lyophilized powder
SAB1303162
ANTI-ANGPT2 (C-TERM) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB2700228
Anti-ANGPT2 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2700229
Anti-ANGPT2 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2700230
Anti-ANGPT2 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB3500709
Anti-ANGPT2 antibody produced in rabbit, affinity isolated antibody
SAB2502065
Anti-S100A8 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB1406407
Anti-S100A8 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA024372
Anti-S100A8 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1411218
Anti-S100A8 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB1411219
Anti-S100A8 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB2500903
Anti-S100A9 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
HPA004193
Anti-S100A9 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1411220
Anti-S100A9 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB2104737
Anti-S100A9, (N-terminal) antibody produced in rabbit, affinity isolated antibody
WH0006279M1
Monoclonal Anti-S100A8 antibody produced in mouse, clone 2H2, purified immunoglobulin, buffered aqueous solution
SAB1404347
Monoclonal Anti-S100A8 antibody produced in mouse, clone 1B3, purified immunoglobulin, buffered aqueous solution
SAB1404348
Monoclonal Anti-S100A9 antibody produced in mouse, clone 4G9, purified immunoglobulin, buffered aqueous solution
SAB1409623
Monoclonal Anti-S100A9 antibody produced in mouse, clone 1C22, purified immunoglobulin, buffered aqueous solution