EMAIL THIS PAGE TO A FRIEND
Food additives & contaminants. Part A, Chemistry, analysis, control, exposure & risk assessment
Migration of selected hydrocarbon contaminants into dry semolina and egg pasta packed in direct contact with virgin paperboard and polypropylene film.
PMID 26209063
Abstract
Migration of mineral oil saturated hydrocarbons (MOSH), polyolefin oligomeric saturated hydrocarbons (POSH), and polyalphaolefins (PAO from hot melts) into dry semolina and egg pasta packed in direct contact with virgin paperboard or polypropylene (PP) flexible film was studied. Migration was monitored during shelf life (up to 24 months), through storage in a real supermarket (packs kept on shelves), conditions preventing exchange with the surrounding environment (packs wrapped in aluminium foil), and storage in a warehouse (packs inside of the transport box of corrugated board). Semolina pasta packed in virgin paperboard (without hot melts) had a MOSH content lower than 1.0 mg kg(-1). An increasing contamination with PAO belonging to the adhesives used to close the boxes was detected in egg pasta, wrapped in aluminium (1.5 and 5 mg kg(-1) after 3 and 24 months, respectively). An environmental contribution to total hydrocarbon contamination was observed in egg pasta kept on shelves that, after 3 and 24 months, showed levels of PAO/MOSH < C25 around 3 and 10 mg kg(-1), respectively. The migration of POSH from PP film into egg pasta wrapped in aluminium was around 0.6 mg kg(-1) after 3 months of contact and reached 1.7 mg kg(-1) after 24 months of contact. After 9 months of contact, semolina pasta packed in PP film and stored in the transport box showed that some MOSH migrated into the pasta from the board of the transport box (through the plastic film).