EMAIL THIS PAGE TO A FRIEND
Histochemistry and cell biology
Impaired extracellular matrix structure resulting from malnutrition in ovariectomized mature rats.
PMID 26210855
Abstract
Bone loss is a symptom related to disease and age, which reflects on bone cells and ECM. Discrepant regulation affects cell proliferation and ECM localization. Rat model of osteoporosis (OVX) was investigated against control rats (Sham) at young and old ages. Biophysical, histological and molecular techniques were implemented to examine the underlying cellular and extracellular matrix changes and to assess the mechanisms contributing to bone loss in the context of aging and the widely used osteoporotic models in rats. Bone loss exhibited a compromised function of bone cells and infiltration of adipocytes into bone marrow. However, the expression of genes regulating collagen catabolic process and adipogenesis was chronologically shifted in diseased bone in comparison with aged bone. The data showed the involvement of Wnt signaling inhibition in adipogenesis and bone loss due to over-expression of SOST in both diseased and aged bone. Further, in the OVX animals, an integrin-mediated ERK activation indicated the role of MAPK in osteoblastogenesis and adipogenesis. The increased PTH levels due to calcium and estrogen deficiency activated osteoblastogenesis. Thusly, RANKL-mediated osteoclastogenesis was initiated. Interestingly, the data show the role of MEPE regulating osteoclast-mediated resorption at late stages in osteoporotic bone. The interplay between ECM and bone cells change tissue microstructure and properties. The involvement of Wnt and MAPK pathways in activating cell proliferation has intriguing similarities to oncogenesis and myeloma. The study indicates the importance of targeting both pathways simultaneously to remedy metabolic bone diseases and age-related bone loss.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A0843
Aluminum sulfate, meets USP testing specifications
Al2O12S3 · xH2O
202614
Aluminum sulfate, 99.99% trace metals basis
Al2O12S3
HPA003157
Anti-BGN antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2104387
Anti-BGN antibody produced in rabbit, affinity isolated antibody
SAB1411228
Anti-BGN antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB1305018
ANTI-CA2 (N-TERM) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
HPA001550
Anti-CA2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2700301
Anti-CA2 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2107491
Anti-CA2 antibody produced in rabbit, affinity isolated antibody
SAB2107492
Anti-CA2 antibody produced in rabbit, affinity isolated antibody
SAB2900220
Anti-ITGB1 (AB1) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2900236
Anti-ITGB1 (AB2) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2107342
Anti-ITGB1 antibody produced in rabbit, affinity isolated antibody
SAB1304508
ANTI-MEPE (N-TERM) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
HPA038004
Anti-MEPE antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA038003
Anti-MEPE antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2106404
Anti-MEPE antibody produced in rabbit, affinity isolated antibody
SAB4501901
Anti-MMP-14, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4501902
Anti-MMP14 (Cleaved-Tyr112) antibody produced in rabbit, affinity isolated antibody
GW21125 Anti-MMP14 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
GW21017 Anti-Tenascin antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
HPA004823
Anti-TNC antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
WH0000633M1
Monoclonal Anti-BGN antibody produced in mouse, clone 4E1-1G7, purified immunoglobulin, buffered aqueous solution
SAB1403603
Monoclonal Anti-BGN antibody produced in mouse, clone 2E6-D1, purified immunoglobulin, buffered aqueous solution
SAB4700394 Monoclonal Anti-CD29 antibody produced in mouse, clone MEM-101A, purified immunoglobulin, buffered aqueous solution
T2551 Monoclonal Anti-Tenascin antibody produced in mouse, clone BC-24, ascites fluid
T3413 Monoclonal Anti-Tenascin antibody produced in rat, clone MTn-12, ascites fluid
WH0003371M1
Monoclonal Anti-TNC antibody produced in mouse, clone 3B4, purified immunoglobulin, buffered aqueous solution
79299
Sodium tartrate dibasic solution, BioUltra, 1.5 M in H2O (colorless solution at 20 °C)
C4H4Na2O6