EMAIL THIS PAGE TO A FRIEND
The Journal of antibiotics
Studies on free radical scavenging substances from microorganisms. I. Carazostatin, a new free radical scavenger produced by Streptomyces chromofuscus DC 118.
PMID 2621170