EMAIL THIS PAGE TO A FRIEND
Cancer science
Hematopoietic pre-B cell leukemia transcription factor interacting protein is overexpressed in gastric cancer and promotes gastric cancer cell proliferation, migration, and invasion.
PMID 26211905
Abstract
Hematopoietic pre-B cell leukemia transcription factor interacting protein (HPIP) has been shown to play an important role in the development and progression of some cancers. However, the role of HPIP in gastric cancer (GC) is unclear. Here, we show that HPIP is upregulated in most GC patients and promotes GC cell proliferation, migration, and invasion. In GC patients, HPIP positively associates with tumor size and nodal metastasis, and negatively associates with tumor differentiation. Hematopoietic pre-B cell leukemia transcription factor interacting protein increases GC cell proliferation through activation of G1 /S and G2 /M cell cycle transitions, accompanied by a marked increase of the positive cell cycle regulators, including cyclin D1, cyclin A, and cyclin B1. Hematopoietic pre-B cell leukemia transcription factor interacting protein enhances GC cell migration and invasion, and modulates epithelial-mesenchymal transition, which plays a key role in cancer cell migration and invasion. These data underscore the critical role of HPIP in GC cell proliferation and progression and suggest that HPIP inhibition may be a useful therapeutic strategy for GC treatment.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB3500308
Anti-Fn14 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB1303493
ANTI-HCFC1R1 (N-TERM) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB2104280
Anti-HCFC1R1 antibody produced in rabbit, affinity isolated antibody
HPA059647
Anti-HCFC1R1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1407783
Anti-PBXIP1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA006949
Anti-PBXIP1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2101733
Anti-PBXIP1 antibody produced in rabbit, affinity isolated antibody
SAB1409412
Anti-TNFRSF12A antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA007853
Anti-TNFRSF12A antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2106931
Anti-TNFRSF12A antibody produced in rabbit, affinity isolated antibody
SAB1412864
Monoclonal Anti-PBXIP1 antibody produced in mouse, clone 7E1, purified immunoglobulin, 1 mg/mL
WH0051330M1 Monoclonal Anti-TNFRSF12A antibody produced in mouse, clone 1B6, purified immunoglobulin, buffered aqueous solution