EMAIL THIS PAGE TO A FRIEND
Mutation research. Genetic toxicology and environmental mutagenesis
Combination comet/micronucleus assay validation performed by BioReliance under the JaCVAM initiative.
PMID 26212297
Abstract
In the international validation study of the in vivo rat alkaline comet assay (comet assay), the Japanese Center for the Validation of Alternative Methods (JaCVAM) provided three coded chemicals to BioReliance, 1,3-dichloropropene, ethionamide and busulfan, to be tested in a combined in vivo comet/micronucleus assay. Induction of DNA damage (comet) in liver, stomach and jejunum (1,3-dichloropropene only) cells, and induction of MNPCEs in bone marrow, were examined in male Sprague-Dawley (Hsd:SD) rats following oral administration of the test chemical for three consecutive days. A dose range finding (DRF) test was performed with each chemical to determine the maximum tolerated dose (MTD). Based on the results of the DRF test; 1,3-dichloropropene was tested at 50, 100 and 200 mg/kg/day; ethionamide was tested at 125, 250 and 500 mg/kg/day, and busulfan was tested at 10, 20 and 40 mg/kg/day. The results indicated that 1,3-dichloropropene induced DNA damage only in liver cells at all three test article doses, while no effects were observed in the stomach and jejunum cells. Additionally, it did not increase MNPCEs in the bone marrow. 1,3-Dichloropropene was concluded to be negative in the MN assay but positive in the comet assay. Ethionamide did not induce DNA damage in liver. However, in stomach, statistically significant decreases (although still within historical range) in % tail DNA at all test article doses compared to the vehicle control were observed. There was no increase in MNPCEs in the bone marrow. Thus, ethionamide was concluded to be negative in the comet/MN combined assay. Busulfan did not induce DNA damage in any of the organs tested (liver and stomach) but it did induce a significant increase in MNPCEs in the bone marrow. Busulfan was concluded to be negative in the comet assay but positive in the MN assay.