EMAIL THIS PAGE TO A FRIEND
Radiation oncology (London, England)
Increased SHP-1 expression results in radioresistance, inhibition of cellular senescence, and cell cycle redistribution in nasopharyngeal carcinoma cells.
PMID 26215037
Abstract
Radioresistance is the main limit to the efficacy of radiotherapy in nasopharyngeal carcinoma (NPC). SHP-1 is involved in cancer progression, but its role in radioresistance and senescence of NPC is not well understood. This study aimed to assess the role of SHP-1 in the radioresistance and senescence of NPC cells. SHP-1 was knocked-down and overexpressed in CNE-1 and CNE-2 cells using lentiviruses. Cells were irradiated to observe their radiosensitivity by colony forming assay. BrdU incorporation assay and flow cytometry were used to monitor cell cycle. A β-galactosidase assay was used to assess senescence. Western blot was used to assess SHP-1, p21, p53, pRb, Rb, H3K9Me3, HP1γ, CDK4, cyclin D1, cyclin E, and p16 protein expressions. Compared with CNE-1-scramble shRNA cells, SHP-1 downregulation resulted in increased senescence (+107%, P < 0.001), increased radiosensitivity, higher proportion of cells in G0/G1 (+33%, P < 0.001), decreased expressions of CDK4 (-44%, P < 0.001), cyclin D1 (-41%, P = 0.001), cyclin E (-97%, P < 0.001), Rb (-79%, P < 0.001), and pRb (-76%, P = 0.001), and increased expression of p16 (+120%, P = 0.02). Furthermore, SHP-1 overexpression resulted in radioresistance, inhibition of cellular senescence, and cell cycle arrest in the S phase. Levels of p53 and p21 were unchanged in both cell lines (all P > 0.05). SHP-1 has a critical role in radioresistance, cell cycle progression, and senescence of NPC cells. Down-regulating SHP-1 may be a promising therapeutic approach for treating patients with NPC.