EMAIL THIS PAGE TO A FRIEND
Dalton transactions (Cambridge, England : 2003)
Covalent bridging of surface functionalized Fe3O4 and YPO4:Eu nanostructures for simultaneous imaging and therapy.
PMID 26215789
Abstract
Magnetic luminescent hybrid nanostructures (MLHN) have received a great deal of attention due to their potential biomedical applications such as thermal therapy, magnetic resonance imaging, drug delivery and intracellular imaging. We report the development of bifunctional Fe3O4 decorated YPO4:Eu hybrid nanostructures by covalent bridging of carboxyl PEGylated Fe3O4 and amine functionalized YPO4:Eu particles. The surface functionalization of individual nanoparticulates as well as their successful conjugation was evident from Fourier transform infrared (FTIR) spectroscopy, dynamic light scattering (DLS), zeta-potential and transmission electron microscopy (TEM) studies. X-ray diffraction (XRD) analysis reveals the formation of highly crystalline hybrid nanostructures. TEM micrographs clearly show the binding/anchoring of 10 nm Fe3O4 nanoparticles onto the surface of 100-150 nm rice grain shaped YPO4:Eu nanostructures. These MLHN show good colloidal stability, magnetic field responsivity and self-heating capacity under an external AC magnetic field. The induction heating studies confirmed localized heating of MLHN under an AC magnetic field with a high specific absorption rate. Photoluminescence spectroscopy and fluorescence microscopy results show optical imaging capability of MLHN. Furthermore, successful internalization of these MLHN in the cells and their cellular imaging ability are confirmed from confocal microscopy imaging. Specifically, the hybrid nanostructure provides an excellent platform to integrate luminescent and magnetic materials into one single entity that can be used as a potential tool for hyperthermia treatment of cancer and cellular imaging.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A3648
(3-Aminopropyl)triethoxysilane, ≥98%
C9H23NO3Si
440140
(3-Aminopropyl)triethoxysilane, 99%
C9H23NO3Si
706493
(3-Aminopropyl)triethoxysilane, packaged for use in deposition systems, ≥98%
C9H23NO3Si
741442
(3-Aminopropyl)triethoxysilane, ≥98.0%
C9H23NO3Si
294993
Ammonia, anhydrous, ≥99.98%
H3N
779423
Ammonia solution, 4 M in methanol
H3N
718939
Ammonia solution, 0.4 M in THF
H3N
608777
Ammonia-14N, 99.99 atom % 14N
H314N
143073
Diglycolic acid, 98%
C4H6O5
380024
Iron(II) chloride tetrahydrate, 99.99% trace metals basis
Cl2Fe · 4H2O
130672
N-Hydroxysuccinimide, 98%
C4H5NO3
225711
Nitric acid, 70%, purified by redistillation, ≥99.999% trace metals basis
HNO3
40230
Nitric acid, semiconductor grade PURANAL (Honeywell 17818), ≥65%
HNO3
40222
Nitric acid, semiconductor grade PURANAL (Honeywell 17952), ≥69%
HNO3
40280
Nitric acid, semiconductor grade MOS PURANAL (Honeywell 17920), ≥69%
HNO3
695041
Nitric acid, ACS reagent, ≥90.0%
HNO3
608769
Nitric-14N acid solution, ~10 N in H2O, 99.99 atom % 14N
H14NO3
333859
Tetraethyl orthosilicate, 99.999% trace metals basis
C8H20O4Si
86578
Tetraethyl orthosilicate, ≥99.0% (GC)
C8H20O4Si
131903
Tetraethyl orthosilicate, reagent grade, 98%
C8H20O4Si
759414
Tetraethyl orthosilicate, packaged for use in deposition systems
C8H20O4Si
262994
Yttrium, chips, 99.9% trace rare earth metals basis
Y
451347
Yttrium, dendritic pieces, 99.9% trace metals basis
Y
693642
Yttrium, foil, thickness 1 mm, 99.8% trace rare earth metals basis
Y
261319
Yttrium, ingot, 99.9% trace rare earth metals basis
Y
261327
Yttrium, powder, −40 mesh, 99.5% trace rare earth metals basis
Y
GF09119279
Yttrium, foil, 25x25mm, thickness 0.15mm, as rolled, 99%
Y
GF77674719
Yttrium, wire reel, 0.2m, diameter 0.5mm, 99.9%
Y
GF11301398
Yttrium, rod, 50mm, diameter 6.35mm, cast, 99%
Y
GF86394156
Yttrium, wire reel, 0.2m, diameter 1.0mm, 99.9%
Y