EMAIL THIS PAGE TO A FRIEND
Pain physician
Intrathecal Baclofen in Spinal Spasticity: Frequency and Severity of Withdrawal Syndrome.
PMID 26218954
Abstract
Intrathecal baclofen (ITB) delivered by programmable pump devices represents an important modality for long-term treatment of severe spinal spasticity. One of the serious adverse events is a withdrawal syndrome after sudden interruption of ITB delivery. In this study, we analyzed the frequency and severity of this complication. Treatment recommendations follow. Case study. Academic medical center. A total of 54 ITB pumps were successfully implanted in 39 patients with severe intractable spasticity (24 with spinal cord injury, 15 with multiple sclerosis, 24 men, age range 21-59 years). Eight patients developed a withdrawal syndrome on total a daily dose of ITB between 90-420 μg/day. Seven patients had catheter-related complications. In one patient, pump failure was observed due to its corrosion. Within the group, baclofen withdrawal syndrome occurred once in 20.1 pump-years counted out of 160.4 pump-years of ITB treatment. Small sample size. ITB withdrawal syndrome is a rare but life-threatening event and prompt diagnosis before treatment initiation is critical. The reported events were mostly mild due to the acute treatment regime and probably due to a lower dose of ITB. A prerequisite for successful ITB treatment is a deep knowledge of complications and their prompt management in the hands of a multidisciplinary team in specialized centers.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B5399
(±)-Baclofen, ≥98% (TLC), solid
C10H12ClNO2