EMAIL THIS PAGE TO A FRIEND
Journal of Crohn's & colitis
Cape Gooseberry [Physalis peruviana L.] Calyces Ameliorate TNBS Acid-induced Colitis in Rats.
PMID 26221001
Abstract
Physalis peruviana [cape gooseberry] is highly appreciated for its commercial value. The Colombian ecotype is in great demand in the international market, particularly for the unique morphological characteristics of the calyx, which has extended use as a traditional herbal remedy in Colombia because of its anti-inflammatory properties. In this work, the anti-inflammatory activity of the total ethereal extract of Physalis peruviana calyces was evaluated in preventive and therapeutic protocols in a TNBS acid-induced colitis rat model. Colitis was induced by intrarectal administration of TNBS. An evaluation of macroscopic and histopathological parameters in colonic tissue was performed, along with the determination of myeloperoxidase enzyme activity, cytokine levels and gene expression. Additionally, effects on nitric oxide release by lipopolysaccharide [LPS]-stimulated RAW264.7 macrophages and the scavenging activity of DPPH and ABTS free radicals were determined. The treatment with the Physalis peruviana extract produced a significant improvement in the colonic tissue at both macroscopic and histological levels. IL-1β and TNF-α production was reduced by the extract in both experimental approaches. The groups treated with Physalis peruviana showed a tendency to MUC2 up-regulation and down-regulation of COX-2, iNOS, NLRP3, IL-1β, IL-6 and IL-10 expression. Nitric oxide release in RAW264.7 macrophages was significantly inhibited. The Physalis peruviana extract showed intestinal anti-inflammatory activity in the TNBS-induced colitis model, placing this species' calyx, a natural derivative, as a promising source of metabolites that could be used in treatment for inflammatory bowel disease.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
238813
(±)-6-Hydroxy-2,5,7,8-tetramethylchromane-2-carboxylic acid, 97%
C14H18O4
281689
1,1-Diphenyl-2-picrylhydrazine, 97%
C18H13N5O6
D9132
2,2-Diphenyl-1-picrylhydrazyl
C18H12N5O6
C7559
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
CHCl3
372978
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
CHCl3
288306
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
CHCl3
439142
Chloroform, for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
CHCl3
437581
Chloroform, ACS reagent, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
CHCl3
480150
Chloroform, ACS reagent, ≥99.8%, contains amylenes as stabilizer
CHCl3
270636
Chloroform, for HPLC, ≥99.8%
CHCl3
48520-U
Chloroform, analytical standard
CHCl3
05-3433
Chloroform, for residue analysis, suitable for 5000 per JIS
CHCl3
05-3450
Chloroform, for HPLC
CHCl3
P2297
Picrylsulfonic acid solution, 5 % (w/v) in H2O, BioReagent, suitable for determination of primary amines
C6H3N3O9S
92823
Picrylsulfonic acid solution, ~1 M in H2O
C6H3N3O9S
P5592
Potassium peroxodisulfate, BioXtra, ≥99.0%
K2O8S2
379824
Potassium persulfate, 99.99% trace metals basis
K2O8S2
216224
Potassium persulfate, ACS reagent, ≥99.0%
K2O8S2
S9251
Sulfanilamide, ≥99%
C6H8N2O2S
33626
Sulfanilamide, puriss. p.a., ≥99% (calc. to the dried substance)
C6H8N2O2S
302643
Trypan Blue, Dye content 60 %
C34H24N6Na4O14S4
T6146
Trypan Blue, powder, BioReagent, suitable for cell culture
C34H24N6Na4O14S4
T8154
Trypan Blue solution, 0.4%, liquid, sterile-filtered, suitable for cell culture
C34H24N6Na4O14S4
224316
Zirconyl chloride octahydrate, reagent grade, 98%
Cl2OZr · 8H2O