EMAIL THIS PAGE TO A FRIEND
Nutrition and cancer
Early Diet and Later Cancer Risk: Prospective Associations of Dietary Patterns During Critical Periods of Childhood with the GH-IGF Axis, Insulin Resistance and Body Fatness in Younger Adulthood.
PMID 26226486
Abstract
Early life, adiposity rebound, and puberty represent critical growth periods when food choices could have long-term relevance for cancer risk. We aimed to relate dietary patterns during these periods to the growth hormone-insulin-like-growth-factor (GH-IGF) axis, insulin resistance, and body fatness in adulthood. Data from the Dortmund Nutritional and Anthropometric Longitudinally Designed (DONALD) Study participants with outcome data at 18-37 years, and ≥2 dietary records during early life (1-2 yr; n = 128), adiposity rebound (4-6 years, n = 179), or puberty (girls 9-14, boys 10-15 yr; n = 213) were used. Dietary patterns at these ages were derived by 1) reduced rank regression (RRR) to explain variation in adult IGF-I, IGF-binding protein-3 (IGFBP-3), homoeostasis model assessment for insulin resistance (HOMA-IR) and fat-mass index; 2) principal component analysis (PCA). Regarding RRR, the patterns "cake/canned fruit/cheese & eggs" (early life), "sweets & dairy" (adiposity rebound) and "high-fat foods" (pubertal boys) were independently associated with higher adult HOMA-IR. Furthermore, the patterns "favorable carbohydrate sources" (early life), "snack & convenience foods" (adiposity rebound), and "traditional & convenience carbohydrates" (pubertal boys) were related to adult IGFBP-3 (P trend < 0.01). PCA identified "healthy" patterns for all periods, but none was associated with the outcomes (P trend > 0.1). In conclusion, dietary patterns during sensitive growth periods may be of long-term relevance for adult insulin resistance and IGFBP-3.
Related Materials