EMAIL THIS PAGE TO A FRIEND
Nutrition journal
Extraction Kinetics of phytochemicals and antioxidant activity during black tea (Camellia sinensis L.) brewing.
PMID 26226943
Abstract
Tea is the most consumed beverage in the world which is second only to water. Tea contains a broad spectrum of active ingredients which are responsible for its health benefits. The composition of constituents extracted to the tea brew depends on the method of preparation for its consumption. The objective of this study was to investigate the extraction kinetics of phenolic compounds, gallic acid, caffeine and catechins and the variation of antioxidant activity with time after tea brew is made. CTC (Crush, Tear, Curl) tea manufactured in Sri Lanka was used in this study. Tea brew was prepared according to the traditional method by adding boiling water to tea leaves. The samples were collected at different time intervals. Total phenolic and flavonoid contents were determined using Folin ciocalteu and aluminium chloride methods respectively. Gallic acid, caffeine, epicatechin, epigallocatechin gallate were quantified by HPLC/UV method. Antioxidant activity was evaluated by DPPH radical scavenging and Ferric Reducing Antioxidant Power (FRAP) assays. Gallic acid, caffeine and catechins were extracted within a very short period. The maximum extractable polyphenols and flavanoids were achieved at 6-8 min after the tea brew is prepared. Polyphenols, flavanoids and epigallocatechin gallate showed a significant correlation (p < 0.001) with the antioxidant activity of tea. The optimum time needed to release tea constituents from CTC tea leaves is 2-8 min after tea is made.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
E3768
(−)-Epigallocatechin, ≥95% (HPLC), from green tea
C15H14O7