EMAIL THIS PAGE TO A FRIEND
Chemistry (Weinheim an der Bergstrasse, Germany)
Facile Microwave-Assisted Solid-Phase Synthesis of Highly Fluorescent Nitrogen-Sulfur-Codoped Carbon Quantum Dots for Cellular Imaging Applications.
PMID 26227302
Abstract
Carbon quantum dots (CQDs) have recently attracted significant attention for both their fundamental science and technological applications as a new class of fluorescent zero-dimensional nanomaterials with a size below 10 nm. However, the reported methods of synthesis were generally less suitable for the large-scale production of the CQDs with high-fluorescent quantum yield (QY). In the paper, a novel one-pot microwave-assisted drying synthesis approach was presented to prepare CQDs with high QY of 61.3 % for the first time. The production yield of CQDs was 35±3 % in weight. The as-prepared CQDs were characterized by various techniques such as TEM, AFM, XRD, XPS, FTIR spectroscopy, UV/Vis absorption spectroscopy, and fluorescence spectroscopy. The results showed that the high QY of CQDs was largely attributed to the dual doping of nitrogen and sulphur into CQDs. Such CQDs were then used as live-cell imaging reagents due to their high QY, good water dispersibility, fine biocompatibility, high photostability, and low cytotoxicity.