EMAIL THIS PAGE TO A FRIEND
World journal of gastroenterology
Association of hepatocyte-derived growth factor receptor/caudal type homeobox 2 co-expression with mucosal regeneration in active ulcerative colitis.
PMID 26229399
Abstract
To characterize the regeneration-associated stem cell-related phenotype of hepatocyte-derived growth factor receptor (HGFR)-expressing cells in active ulcerative colitis (UC). On the whole 38 peripheral blood samples and 38 colonic biopsy samples from 18 patients with histologically proven active UC and 20 healthy control subjects were collected. After preparing tissue microarrays and blood smears HGFR, caudal type homeobox 2 (CDX2), prominin-1 (CD133) and Musashi-1 conventional and double fluorescent immunolabelings were performed. Immunostained samples were digitalized using high-resolution Mirax Desk instrument, and analyzed with the Mirax TMA Module software. For semiquantitative counting of immunopositive lamina propria (LP) cells 5 fields of view were counted at magnification × 200 in each sample core, then mean ± SD were determined. In case of peripheral blood smears, 30 fields of view with 100 μm diameter were evaluated in every sample and the number of immunopositive cells (mean ± SD) was determined. Using 337 nm UVA Laser MicroDissection system at least 5000 subepithelial cells from the lamina propria were collected. Gene expression analysis of HGFR, CDX2, CD133, leucine-rich repeat-containing G-protein coupled receptor 5 (Lgr5), Musashi-1 and cytokeratin 20 (CK20) were performed in both laser-microdisscted samples and blood samples by using real time reverse transcription polymerase chain reaction (RT-PCR). By performing conventional and double fluorescent immunolabelings confirmed by RT-PCR, higher number of HGFR (blood: 6.7 ± 1.22 vs 38.5 ± 3.18; LP: 2.25 ± 0.85 vs 9.22 ± 0.65; P < 0.05), CDX2 (blood: 0 vs 0.94 ± 0.64; LP: 0.75 ± 0.55 vs 2.11 ± 0.75; P < 0.05), CD133 (blood: 1.1 ± 0.72 vs 8.3 ± 1.08; LP: 11.1 ± 0.85 vs 26.28 ± 1.71; P < 0.05) and Musashi-1 (blood and LP: 0 vs scattered) positive cells were detected in blood and lamina propria of UC samples as compared to controls. HGFR/CDX2 (blood: 0 vs 1 ± 0.59; LP: 0.8 ± 0.69 vs 2.06 ± 0.72, P < 0.05) and Musashi-1/CDX2 (blood and LP: 0 vs scattered) co-expressions were found in blood and lamina propria of UC samples. HGFR/CD133 and CD133/CDX2 co-expressions appeared only in UC lamina propria samples. CDX2, Lgr5 and Musashi-1 expressions in UC blood samples were not accompanied by CK20 mRNA expression. In active UC, a portion of circulating HGFR-expressing cells are committed to the epithelial lineage, and may participate in mucosal regeneration by undergoing mesenchymal-to-epithelial transition.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB2500223
Anti-CDX2 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
AV31476
Anti-CDX2 antibody produced in rabbit, affinity isolated antibody
SAB3500187
Anti-CDX2 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
F3008 Anti-Mouse IgG (whole molecule)–FITC antibody produced in sheep, affinity isolated antibody, buffered aqueous solution
SAB1406144
Anti-MSI1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB1410797
Anti-MSI1 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
M3571 Anti-Musashi-1 antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
AP132B
Goat Anti-Rabbit IgG Antibody, biotin-SP conjugate, 2 mL, Goat anti-Rabbit IgG Antibody, biotin-SP conjugate detects level of Rabbit IgG & has been published & validated for use in ELISA & WB.
WH0001045M1
Monoclonal Anti-CDX2 antibody produced in mouse, clone 1C7, purified immunoglobulin, buffered aqueous solution
SAB4100260
Monoclonal Anti-MSI1 antibody produced in mouse, clone MSI1-A230, culture supernatant
SAB4100261
Monoclonal Anti-MSI1 antibody produced in mouse, clone MSI1-B230, culture supernatant
SAB1409328
Monoclonal Anti-MSI1 antibody produced in mouse, clone 3E4, purified immunoglobulin, buffered aqueous solution
SAB1409329
Monoclonal Anti-MSI1 antibody produced in mouse, clone 4C7, purified immunoglobulin, buffered aqueous solution
SAB1409330
Monoclonal Anti-MSI1 antibody produced in mouse, clone 1C3, purified immunoglobulin, buffered aqueous solution
SAB1404084
Monoclonal Anti-MSI1, (N-terminal) antibody produced in mouse, clone 1B11, purified immunoglobulin, buffered aqueous solution
SAB1402276
Monoclonal Anti-MSI1, (N-terminal) antibody produced in mouse, clone 3F2, purified immunoglobulin, buffered aqueous solution