EMAIL THIS PAGE TO A FRIEND
Cell & bioscience
Exogenous hydrogen sulfide restores cardioprotection of ischemic post-conditioning via inhibition of mPTP opening in the aging cardiomyocytes.
PMID 26229588
Abstract
The physiological and pathological roles of hydrogen sulfide (H2S) in the regulation of cardiovascular functions have been recognized. H2S protects against the hypoxia/reoxygenation (H/R)-induced injury and apoptosis of cardiomyocytes, and ischemic post-conditioning (PC) plays an important role in cardioprotection from H/R injury in neonatal cardiomyocytes but not in aging cardiomyocytes. Whether H2S is involved in the recovery of PC-induced cardioprotection in aging cardiomyocytes is unclear. In the present study, we found that both H/R and PC decreased cystathionine-γ-lyase (CSE) expression and the production rate of H2S. Supplementation of NaHS protected against H/R-induced apoptosis, the expression of cleaved caspase-3 and cleaved caspase-9, the release of cytochrome c (Cyt c), and mPTP opening. The addition of NaHS also counteracted the reduction of cell viability caused by H/R and increased the phosphorylation of ERK1/2, PI3K, Akt, GSK-3β and mitochondrial membrane potential. Additionally, NaHS increased Bcl-2 expression, promoted PKC-ε translocation to the cell membrane, and activated mitochondrial ATP-sensitive K channels (mitoKATP). PC alone did not provide cardioprotection in H/R-treated aging cardiomyocytes, which was significantly restored by the supplementation of NaHS. In conclusion, our results suggest that exogenous H2S restores PC-induced cardioprotection via the inhibition of mPTP opening by the activation of the ERK1/2-GSK-3β, PI3K-Akt-GSK-3β and PKC-ε-mitoKATP pathways in aging cardiomyocytes. These findings provide a novel target for the treatment of aging ischemic cardiomyopathy.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB1405673
Anti-CTH antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
AV46068
Anti-CTH antibody produced in rabbit, IgG fraction of antiserum
HPA021591
Anti-CTH antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab1
HPA023300
Anti-CTH antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab2
SAB2100501
Anti-CTH antibody produced in rabbit, affinity isolated antibody
SAB4501330
Anti-GSK3 antibody produced in rabbit, affinity isolated antibody
SAB4300287
Anti-GSK3B (Ab-9) antibody produced in rabbit, affinity isolated antibody
SAB2100978
Anti-GSK3B antibody produced in rabbit, affinity isolated antibody
HPA028017
Anti-GSK3B antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
C2932
Chelerythrine chloride, ≥95% (TLC), powder
C21H18ClNO4
SAB1403711
Monoclonal Anti-CTH antibody produced in mouse, clone S51, purified immunoglobulin, buffered aqueous solution
WH0002932M4
Monoclonal Anti-GSK3B antibody produced in mouse, clone 1H3, purified immunoglobulin, buffered aqueous solution
SAB5300175 Monoclonal Anti-GSK3B antibody produced in mouse, clone 3D10, ascites fluid