PloS one
Abnormally Low or High Ankle-Brachial Index Is Associated with Proliferative Diabetic Retinopathy in Type 2 Diabetic Mellitus Patients.
PMID 26230390
Abstract
Although some studies have reported that low ankle-brachial index (ABI) is associated with diabetic retinopathy (DR) in diabetic patients, it remains controversial as to which stage of DR. The aim of this study is to assess whether peripheral artery disease (PAD), indicated by abnormally low or high ABI, is associated with different stages of DR in patients with type 2 diabetes mellitus (DM), and further evaluate the risk factors. A total of 2001 (858 men and 1143 women) patients with type 2 DM who underwent ABI measurement in an outpatient clinic were enrolled. PAD was defined as ABI < 0.9 or ≧ 1.3 in either leg. DR was classified as non-DR, nonproliferative DR and proliferative DR stages. The clinical data were analyzed and the risk factors for abnormal ABI were determined by multivariate logistic regression analysis. The prevalence of ABI < 0.9 or ≧ 1.3 was 3.0%. Multivariate forward logistic regression analysis identified proliferative DR (vs. non-DR) was associated with abnormal ABI (odds ratio, 1.718; 95% confidence interval, 1.152 to 2.562; p = 0.008), but nonproliferative DR was not. Furthermore, the presence of coronary artery disease, cerebrovascular disease, declining renal function and patients without diuretics use were associated with abnormal ABI in patients with proliferative DR. Our study in patients of type 2 DM demonstrated that PAD was associated with proliferative DR. We emphasize the recommendation of performing the ABI test in this population at risk.