EMAIL THIS PAGE TO A FRIEND
Analytica chimica acta
Poly(N-vinylimidazole/ethylene glycol dimethacrylate) for the purification and isolation of phenolic acids.
PMID 26231906
Abstract
In this study we report the novel polymeric resin poly(N-vinyl imidazole/ethylene glycol dimethacrylate) for the purification and isolation of phenolic acids. The monomer to crosslinker ratio and the porogen composition were optimized for isolating phenolic acids diluted in acetonitrile at normal phase chromatography conditions, first. Acetonitrile serves as polar, aprotic solvent, dissolving phenolic acids but not interrupting interactions with the stationary phase due to the approved Hansen solubility parameters. The optimized resin demonstrated high loading capacities and adsorption abilities particularly for phenolic acids in both, acetonitrile and aqueous solutions. The adsorption behavior of aqueous standards can be attributed to ion exchange effects due to electrostatic interactions between protonated imidazole residues and deprotonated phenolic acids. Furthermore, adsorption experiments and subsequent curve fittings provide information of maximum loading capacities of single standards according to the Langmuir adsorption model. Recovery studies of the optimized polymer in the normal-phase and ion-exchange mode illustrate the powerful isolation properties for phenolic acids and are comparable or even better than typical, commercially available solid phase extraction materials. In order to prove the applicability, a highly complex extract of rosemary leaves was purified by poly(N-vinyl imidazole/ethylene glycol dimethacrylate) and the isolated compounds were identified using UHPLC-qTOF-MS.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
240559
1,4-Butanediol, ReagentPlus®, ≥99%
C4H10O2
493732
1,4-Butanediol, ReagentPlus®, 99%
C4H10O2
279544
1-Propanol, anhydrous, 99.7%
C3H8O
W292818
1-Propanol, ≥99%, FG
C3H8O
W292826
1-Propanol, natural, ≥98%, FG
C3H8O
24-6080
1-Propanol, SAJ first grade, ≥99.0%
C3H8O
24-6070
1-Propanol, JIS special grade
C3H8O
235466
1-Vinylimidazole, ≥99%
C5H6N2
360066
2,2,4-Trimethylpentane, anhydrous, 99.8%
C8H18
15-2200
2,2,4-Trimethylpentane, SAJ first grade, ≥98.0%
C8H18
15-2190
2,2,4-Trimethylpentane, JIS special grade
C8H18
15-2211
2,2,4-Trimethylpentane, for HPLC
C8H18
126209
2,3-Dihydroxybenzoic acid, 99%
C7H6O4
102601
3,4-Dihydroxyhydrocinnamic acid, 98%
C9H10O4
C0625
Caffeic acid, ≥98.0% (HPLC)
C9H8O4
60020
Caffeic acid, purum, ≥95.0% (HPLC)
C9H8O4
C3878
Chlorogenic acid, ≥95% (titration)
C16H18O9
105899
Cyclohexanol, ReagentPlus®, 99%
C6H12O
57180
Indo 1, for fluorescence, ≥90% (HPLC)
C32H26K5N3O12
W261718
Lauryl alcohol, ≥98%, FG
C12H26O
536954
Rosmarinic acid, 96%
C18H16O8
R4033
Rosmarinic acid, ≥98% (HPLC), from Rosemarinus officinalis L.
C18H16O8
403040
Tannic acid, ACS reagent
C76H52O46
T0200
Tannic acid, Source: Chinese natural gall nuts
C76H52O46
W304204
Tannic acid
C76H52O46
W518301
trans-Ferulic acid, ≥99%
C10H10O4
128708
trans-Ferulic acid, 99%
C10H10O4