EMAIL THIS PAGE TO A FRIEND
American journal of physiology. Heart and circulatory physiology
Differential effects of octanoate and heptanoate on myocardial metabolism during extracorporeal membrane oxygenation in an infant swine model.
PMID 26232235
Abstract
Nutritional energy support during extracorporeal membrane oxygenation (ECMO) should promote successful myocardial adaptation and eventual weaning from the ECMO circuit. Fatty acids (FAs) are a major myocardial energy source, and medium-chain FAs (MCFAs) are easily taken up by cell and mitochondria without membrane transporters. Odd-numbered MCFAs supply carbons to the citric acid cycle (CAC) via anaplerotic propionyl-CoA as well as acetyl-CoA, the predominant β-oxidation product for even-numbered MCFA. Theoretically, this anaplerotic pathway enhances carbon entry into the CAC, and provides superior energy state and preservation of protein synthesis. We tested this hypothesis in an immature swine model undergoing ECMO. Fifteen male Yorkshire pigs (26-45 days old) with 8-h ECMO received either normal saline, heptanoate (odd-numbered MCFA), or octanoate (even-numbered MCFA) at 2.3 μmol·kg body wt(-1)·min(-1) as MCFAs systemically during ECMO (n = 5/group). The 13-carbon ((13)C)-labeled substrates ([2-(13)C]lactate, [5,6,7-(13)C3]heptanoate, and [U-(13)C6]leucine) were systemically infused as metabolic markers for the final 60 min before left ventricular tissue extraction. Extracted tissues were analyzed for the (13)C-labeled and absolute concentrations of metabolites by nuclear magnetic resonance and gas chromatography-mass spectrometry. Octanoate produced markedly higher myocardial citrate concentration, and led to a higher [ATP]-to-[ADP] ratio compared with other groups. Unexpectedly, octanoate and heptanoate increased the flux of propionyl-CoA relative to acetyl-CoA into the CAC compared with control. MCFAs promoted increases in leucine oxidation, but were not associated with a difference in protein synthesis rate. In conclusion, octanoate provides energetic advantages to the heart over heptanoate.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
L7875
DL-Leucine, ≥99% (HPLC)
C6H13NO2
190144
Pyridinium chlorochromate, 98%
C5H5N · ClCrO3H