EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry
Dual inhibitors of the dengue and West Nile virus NS2B-NS3 proteases: Synthesis, biological evaluation and docking studies of novel peptide-hybrids.
PMID 26233795
Abstract
Dengue virus (DENV) and West Nile virus (WNV) are mosquito-borne arboviruses responsible for causing acute systemic diseases and severe health conditions in humans. The discovery of therapies capable to prevent infections or treat infected individuals remains an important challenge, since no vaccine or specific efficient treatment could be developed so far. In this context, we present herein the synthesis, characterization, biological evaluation and docking studies of novel peptide-hybrids based on 2,4-thiazolidinedione scaffolds containing non-polar groups. The most promising compound has an IC50 of 0.75 μM against WNV protease, which represents a seventyfold improvement in activity compared to our previously reported compounds. Experimental results and docking studies are in agreement with the hypothesis that a non-polar group in the scaffold is important to obtain interactions between the inhibitors and a hydrophobic pocket in the substrate recognition region of the DENV and WNV NS2B-NS3 serine proteases.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
12804
HBTU, ≥98.0% (T)
C11H16F6N5OP