EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry
Synthesis and biological evaluation of 9-N-oxamyl sialosides as Siglec-7 ligands.
PMID 26234906
Abstract
Siglecs (sialic acid recognizing immunoglobulin like lectins) are a family of lectins with specificity for sialic acid containing carbohydrates. Synthetic sialic acid derivatives with high affinity proved useful to unravel the biological role of the ligand binding domain, although many of their functions in immunity remain unknown. Here we present design, synthesis, affinity evaluation and molecular modeling of novel 9-N-oxamoyl modified sialosides as Siglec-7 ligands.