EMAIL THIS PAGE TO A FRIEND
Chemical & pharmaceutical bulletin
Megastigmane Glucosides and Megastigmanes from the Leaves of Meliosma lepidota ssp. squamulata.
PMID 26235168
Abstract
From the leaves of Meliosma lepidota ssp. squamulata, megastigmane glucosides with spiro-structures and megastigmanes were isolated. Their structures were determined by X-ray crystallographic analyses and spectroscopic investigation. The absolute structures of the megastigmanes were determined by the modified Mosher's method.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
156558
α-Methoxy-α-(trifluoromethyl)phenylacetic acid, 97%
C10H9F3O3