EMAIL THIS PAGE TO A FRIEND
Biological & pharmaceutical bulletin
The 5-Lipoxygenase Inhibitor Zileuton Confers Neuroprotection against Glutamate Oxidative Damage by Inhibiting Ferroptosis.
PMID 26235588
Abstract
5-Lipoxygenase (5-LOX) inhibitors have been shown to be protective in several neurodegenerative disease models; however, the underlying mechanisms remain unclear. We investigated whether 5-LOX inhibitor zileuton conferred direct neuroprotection against glutamate oxidative toxicity by inhibiting ferroptosis, a newly identified iron-dependent programmed cell death. Treatment of HT22 mouse neuronal cell line with glutamate resulted in significant cell death, which was inhibited by zileuton in a dose-dependent manner. Consistently, zileuton decreased glutamate-induced production of reactive oxygen species but did not restore glutamate-induced depletion of glutathione. Moreover, the pan-caspase inhibitor Z-Val-Ala-Asp(OMe)-fluoromethyl ketone (ZVAD-fmk) neither prevented HT22 cell death induced by glutamate nor affected zileuton protection against glutamate oxidative toxicity, suggesting that zileuton did not confer neuroprotection by inhibiting caspase-dependent apoptosis. Interestingly, glutamate-induced HT22 cell death was significantly inhibited by the ferroptosis inhibitor ferrostatin-1. Moreover, zileuton protected HT22 neuronal cells from erastin-induced ferroptosis. However, we did not observe synergic protective effects of zileuton and ferrostatin-1 on glutamate-induced cell death. These results suggested that both the 5-LOX inhibitor zileuton and the ferropotosis inhibitor ferrostatin-1 acted through the same cascade to protect against glutamate oxidative toxicity. In conclusion, our results suggested that zileuton protected neurons from glutamate-induced oxidative stress at least in part by inhibiting ferroptosis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
E7781
Erastin, ≥98% (HPLC)
C30H31ClN4O4