EMAIL THIS PAGE TO A FRIEND
Scientific reports
HAUSP-nucleolin interaction is regulated by p53-Mdm2 complex in response to DNA damage response.
PMID 26238070
Abstract
HAUSP (herpes virus-associated ubiquitin specific protease, known as ubiquitin specific protease 7), one of DUBs, regulates the dynamics of the p53 and Mdm2 network in response to DNA damage by deubiquitinating both p53 and its E3 ubiquitin ligase, Mdm2. Its concerted action increases the level of functional p53 by preventing proteasome-dependent degradation of p53. However, the protein substrates that are targeted by HAUSP to mediate DNA damage responses in the context of the HAUSP-p53-Mdm2 complex are not fully identified. Here, we identified nucleolin as a new substrate for HAUSP by proteomic analysis. Nucleolin has two HAUSP binding sites in its N- and C-terminal regions, and the mutation of HAUSP interacting peptides on nucleolin disrupts their interaction and it leads to the increased level of nucleolin ubiquitination. In addition, HAUSP regulates the stability of nucleolin by removing ubiquitin from nucleolin. Nucleolin exists as a component of the HAUSP-p53-Mdm2 complex, and both Mdm2 and p53 are required for the interaction between HAUSP and nucleolin. Importantly, the irradiation increases the HAUSP-nucleolin interaction, leading to nucleolin stabilization significantly. Taken together, this study reveals a new component of the HAUSP-p53-Mdm2 complex that governs dynamic cellular responses to DNA damage.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB4300601
Anti-MDM2 (Ab-166) antibody produced in rabbit, affinity isolated antibody
SAB2500625
Anti-MDM2 (Isoform) antibody produced in goat, affinity isolated antibody, buffered aqueous solution
GW21160 Anti-MDM2 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB4501849
Anti-MDM2 antibody produced in rabbit, affinity isolated antibody
SAB4501850
Anti-MDM2 antibody produced in rabbit, affinity isolated antibody
SAB4501851
Anti-MDM2, C-Terminal antibody produced in rabbit, affinity isolated antibody
AV40583
Anti-NCL (AB1) antibody produced in rabbit, affinity isolated antibody
AV40584
Anti-NCL (AB2) antibody produced in rabbit, IgG fraction of antiserum
SAB1409341
Anti-NCL antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA023981
Anti-NCL antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1410823
Anti-NCL antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
N2662
Anti-Nucleolin antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB1306667
ANTI-P53 (S15) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB1306713
ANTI-P53 (S315) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB1306623
ANTI-P53 (SUMOYLATION SITE SPECIFIC) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB1306675
ANTI-P53 (T55) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB4503015
Anti-p53 antibody produced in rabbit, affinity isolated antibody
P4235 Anti-p53 antibody produced in sheep, fractionated antiserum, buffered aqueous solution
SAB1306006
ANTI-P53(C-TERMINAL) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB4503001
Anti-p53, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4503003
Anti-p53, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4503010
Anti-p53, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4503011
Anti-p53, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4503012
Anti-p53, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4503013
Anti-p53, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4503019
Anti-p53, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4503021
Anti-p53, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4300365
Anti-TP53 (Ab-18) antibody produced in rabbit, affinity isolated antibody
SAB4300369
Anti-TP53 (Ab-315) antibody produced in rabbit, affinity isolated antibody
SAB4300366
Anti-TP53 (Ab-33) antibody produced in rabbit, affinity isolated antibody